Skip to Content
Merck
All Photos(4)

Key Documents

P0169

Sigma-Aldrich

Palmityl palmitate

≥99%

Synonym(s):

Cetyl palmitate, Hexadecyl hexadecanoate, Palmitic acid palmityl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2(CH2)15CH3
CAS Number:
Molecular Weight:
480.85
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99%

form

powder

mp

55-56 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

InChI

1S/C32H64O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-34-32(33)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3

InChI key

PXDJXZJSCPSGGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paolo Blasi et al.
International journal of pharmaceutics, 419(1-2), 287-295 (2011-08-11)
The aim of this study was to optimize the formulation of lipid nanoparticles (NPs), intended for brain targeting, with the aid of a computer generated experimental design. The high pressure homogenization technique, selected for this purpose, was suitable to formulate
G Lukowski et al.
International journal of pharmaceutics, 196(2), 201-205 (2000-03-04)
Solid lipid nanoparticles (SLN) as alternative intravenous colloidal drug carriers were produced by high pressure homogenisation of the melted lipid cetylpalmitate. The crystallographic properties of the used cetylpalmitate SLN were characterised by small angle X-ray scattering (SAXS) and X-ray diffraction
Susana Martins et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 45(5), 613-623 (2012-01-17)
Physicochemical properties of lipid nanoparticles (LN), such as size, size distribution and surface charge, have a major influence both, on in vitro stability and delivery of the incorporated drug in vivo. With the purpose of understanding how these properties are
Effat Sadat Farboud et al.
International journal of nanomedicine, 6, 611-617 (2011-06-16)
Solid lipid nanoparticles (SLNs) of coenzyme Q10 (CoQ10) were formulated by a high-pressure homogenization method. The best formulation of SLN dispersion consisted of 13% lipid (cetyl palmitate or stearic acid), 8% surfactant (Tween 80 or Tego Care 450), and water.
N Schöler et al.
International journal of pharmaceutics, 196(2), 235-239 (2000-03-04)
In order to investigate the interaction of preserved solid lipid nanoparticles (SLN) with murine peritoneal macrophages (Mpsi), cytotoxicity and proinflammatory effects of two different solid lipid nanoparticles (SLN) preparations consisting of either compritol (CO) or cetyl palmitate (CP) preserved with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service