Skip to Content
Merck
All Photos(1)

Key Documents

P0151

Sigma-Aldrich

Poly(Glu, Tyr) sodium salt

Glu:Tyr (1:1), mol wt 20,000-50,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26
Pricing and availability is not currently available.

form

solid

Quality Level

feed ratio

Glu:Tyr (1:1)

mol wt

20,000-50,000

application(s)

cell analysis

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Poly(glu, tyr) sodium salt is a polyamino acid with applications in cancer research.

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Bruns et al.
Cancer research, 60(11), 2926-2935 (2000-06-13)
We determined whether down-regulation of the epidermal growth factor-receptor (EGF-R) signaling pathway by oral administration of a novel EGF-R tyrosine kinase inhibitor (PKI166) alone or in combination with gemcitabine (administered i.p.) can inhibit growth and metastasis of human pancreatic carcinoma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service