Skip to Content
Merck
All Photos(1)

Key Documents

P0069

Sigma-Aldrich

PRIMA-1

≥98% (HPLC), solid

Synonym(s):

2,2-Bishydroxymethyl-1-aza-bicyclo[2.2.2]octan-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3
CAS Number:
Molecular Weight:
185.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

OCC1(CO)N2CC[C@@H](CC2)C1=O

InChI

1S/C9H15NO3/c11-5-9(6-12)8(13)7-1-3-10(9)4-2-7/h7,11-12H,1-6H2

InChI key

RFBVBRVVOPAAFS-UHFFFAOYSA-N

Related Categories

General description

P53-dependent reactivation and induction of massive apoptosis (PRIMA-1) is a low molecular weight compound and has a role in cancer therapy. PRIMA-1 stimulates cytotoxic effects in human cancer cells. It induces eryptosis.

Biochem/physiol Actions

PRIMA-1 is a selective re-activator of mutant p53 activity in tumor cells, and an inducer of apoptosis and inhibitor of growth of human tumors with mutant p53.
PRIMA-1 is a selective re-activator of mutant p53 activity in tumor cells, and an inducer of apoptosis and inhibitor of growth of human tumors with mutant p53. Mutations in the tumor suppressor p53 take place in >50% tumor cells. PRIMA-1 selectively restores sequence-specific DNA binding and transactivational activity to mutant p53 proteins at μM concentrations. PRIMA-1 works as a re-activator of the apoptotic function of mutant p53 via conformational modulation of function-specific epitopes.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Idogawa et al.
Oncotarget, 5(17), 7540-7548 (2014-10-04)
p53 transduction is a potentially effective cancer therapy but does not result in a good therapeutic response in all human cancers due to resistance to apoptosis. To discover factors that overcome resistance to p53-induced apoptosis, we attempted to identify RNAi
Stimulation of suicidal erythrocyte death by PRIMA-1
Faggio C, et al.
Cellular Physiology and Biochemistry, 35(2), 529-540 (2015)
PRIMA-1 as a cancer therapy restoring mutant p53: A review
Lewis EJ
Bioscience Horizons, 8 (2015)
Federica Eduati et al.
Molecular systems biology, 16(2), e8664-e8664 (2020-02-20)
Mechanistic modeling of signaling pathways mediating patient-specific response to therapy can help to unveil resistance mechanisms and improve therapeutic strategies. Yet, creating such models for patients, in particular for solid malignancies, is challenging. A major hurdle to build these models
Yantao Han et al.
Cell biology international, 41(12), 1296-1306 (2017-08-10)
Vascular smooth muscle cell (VSMC) proliferation is a major contributor to atherosclerosis. This study investigated the inhibitory effects of oleanolic acid (OA) against oxidized low-density lipoprotein (ox-LDL)-induced VSMC proliferation in A7r5 cells and explored underlying molecular mechanism. The cell proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service