Skip to Content
Merck
All Photos(1)

Key Documents

O8889

Sigma-Aldrich

Oleuropein

≥80% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32O13
CAS Number:
Molecular Weight:
540.51
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

description

Synthetic

Quality Level

Assay

≥80% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

room temp

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)[C@@H]1CC(=O)OCCc3ccc(O)c(O)c3

InChI

1S/C25H32O13/c1-3-13-14(9-19(29)35-7-6-12-4-5-16(27)17(28)8-12)15(23(33)34-2)11-36-24(13)38-25-22(32)21(31)20(30)18(10-26)37-25/h3-5,8,11,14,18,20-22,24-28,30-32H,6-7,9-10H2,1-2H3/b13-3+/t14-,18+,20+,21-,22+,24-,25-/m0/s1

InChI key

RFWGABANNQMHMZ-ZCHJGGQASA-N

General description

Oleuropein is one of the main phenolic compounds of olive oil, which gives a bitter and pungent taste to the extra-virgin olive oil. It is a type of secoiridoids and an ester of 3-Hydroxytyrosol with an oleosidic skeleton. Oleuropein is found in the oil and fruits of Olea europaea and also in genera belonging to the Oleaceae family.

Application

Oleuropein has been used:
  • as an anticancer agent to test its effect on different apoptotic pathway regulators in two breast cancer cell lines (MCF7 and MDA-MB-231) in cell viability assay
  • as an antioxidant to test its effect on the hydrogen peroxide (H2O2)-induced premature senescence in human fibroblast cell lines
  • to test its anticancer properties on the cell viability, cisplatin resistance, and apoptosis in two ovarian cell lines (A2780S and A2780/CP cell lines)
  • as a model antioxidant to analyze the quenching activity of 2,2-diphenyl-1-picrylhydrazyl (DPPH) in different solvents

Biochem/physiol Actions

Oleuropein has many pharmacological benefits with its antioxidant, anti-inflammatory, antimicrobial, antiviral, hypoglycemic, hypolipidemic, neuroprotective, and anti-aging effects. It has shown antimycoplasmal activity against Mycoplasma strains in in vitro studies.
Phenolic constituent of olive oil. Contributes to anticancer properties of olive oil. Oleuropein and its metabolite, hydroxytyrosol, inhibit the rate of cancer cell proliferation and
induce cancer cell apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syed Haris Omar
Scientia pharmaceutica, 78(2), 133-154 (2010-12-24)
Olive from Olea europaea is native to the Mediterranean region and, both the oil and the fruit are some of the main components of the Mediterranean diet. The main active constituents of olive oil include oleic acid, phenolic constituents, and
Somayeh Hashemi Sheikhshabani et al.
Life sciences, 278, 119525-119525 (2021-04-25)
Despite many attempts to treat ovarian cancer, 13,940 individuals perish annually due to this disease worldwide. Chemotherapy is the main approach to ovarian cancer treatment, but the development of drug resistance is a major obstacle to the successful treatment. Oleuropein
Pio Maria Furneri et al.
International journal of antimicrobial agents, 20(4), 293-296 (2002-10-19)
The activity of oleuropein, a phenolic glycoside contained in olive oil, was investigated in vitro against Mycoplasma hominis, Mycoplasma fermentans, Mycoplasma pneumoniae and Mycoplasma pirum. Oleuropein inhibited mycoplasmas at concentrations from 20 to 320 mg/l. The MICs of oleuropein to
Samira Asgharzade et al.
European journal of pharmacology, 886, 173509-173509 (2020-09-06)
In spite of advancements in breast cancer therapy, this disease is still one of the significant causes of women fatalities globally. Dysregulation of miRNA plays a pivotal role in the initiation and progression of cancer. Therefore, the administration of herbal
Aderonke Oyetunji et al.
Psychiatry research, 284, 112769-112769 (2020-01-22)
Postpartum stress has been shown to have an association with infant growth, nutrition, bonding, temperament and ultimately childhood mental wellbeing. When persistent, recurring or chronic, it can negatively impact infant outcome, including the subscales of mental wellbeing such as growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service