Skip to Content
Merck
All Photos(1)

Key Documents

NA0150S

Sigma-Aldrich

GenElute HP Plasmid Miniprep Kit

sufficient for 10 preparations

Synonym(s):

GenElute HP Plasmid Kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.52

usage

sufficient for 10 preparations

Quality Level

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the plasmid DNA onto silica in the presence of high salts. Contaminants are then removed by a vacuum or spin wash steps. Finally, the bound plasmid DNA is eluted in water or Tris-EDTA buffer.

The GenElute HP Plasmid Miniprep Kit offers an ultrafast and efficient solution for plasmid preparation from E. coli cultures. This kit combines silica-based membrane technology and the convenience of spin or vacuum format,up to 25 μg of high copy plasmid DNA can be recovered from 1-5 mL of E. coli culture in less than 30 minutes. Note that actual yield and optimum volume of culture to use depend on the plasmid and the culture medium.
An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the plasmid DNA onto silica in the presence of high salts. Contaminants are then removed by a vacuum or spin wash steps. Finally, the bound plasmid DNA is eluted in water or Tris buffer.

The GenElute HP Plasmid Miniprep Kit offers an ultrafast and efficient solution for plasmid preparation from E. coli cultures. This kit combines silica-based membrane technology and the convenience of spin or vacuum format,up to 25 μg of high copy plasmid DNA can be recovered from 1-5 mL of E. coli culture in less than 30 minutes. Note that actual yield and optimum volume of culture to use depend on the plasmid and the culture medium.

Application

The recovered plasmid DNA is ready for immediate use in applications such as restriction enzyme digestion, cloning, PCR, transcription, and sequencing.

Features and Benefits

  • Purified plasmid DNA in less than 30 minutes
  • Up to 25 μg of high-copy plasmid DNA
  • Flexibility of a vacuum or spin format
  • No phenol/chloroform extraction or alcohol precipitation required

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole M I Nivillac et al.
Biochimica et biophysica acta, 1788(10), 2326-2334 (2009-08-25)
Human Equilibrative Nucleoside Transporter 1 (hENT1) is an integral membrane protein that transports nucleosides and analog drugs across cellular membranes. Very little is known about intracellular processing and localization of hENT1. Here we show that disruption of a highly conserved
Katherine Guild et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 67(Pt 9), 1027-1031 (2011-09-10)
Recombinant expression of proteins of interest in Escherichia coli is an important tool in the determination of protein structure. However, lack of expression and insolubility remain significant challenges to the expression and crystallization of these proteins. The SSGCID program uses
Emilia Vuttariello et al.
Molecular biotechnology, 54(3), 954-960 (2013-01-29)
Hereditary cancers account for approximately 10 % of breast and ovarian cancers. Mutations of the BRCA1 and BRCA2 genes, encoding two proteins involved in DNA repair, underlie most cases of such hereditary cancers. Women with BRCA mutations develop breast cancer
Radhika Goenka et al.
The Journal of experimental medicine, 211(1), 45-56 (2013-12-25)
We have assessed the role of B lymphocyte stimulator (BLyS) and its receptors in the germinal center (GC) reaction and affinity maturation. Despite ample BLyS retention on B cells in follicular (FO) regions, the GC microenvironment lacks substantial BLyS. This
Antonietta Carbone et al.
Acta biochimica Polonica, 59(2), 275-278 (2012-06-14)
Supercoiled state corresponds to the active form for plasmid applications. The relaxed circular form of plasmids is often inactive or poorly active. To obtain significant amounts of almost fully supercoiled DNA, we modified the standard protocol of a commercially available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service