Skip to Content
Merck
All Photos(2)

Key Documents

N8028

Sigma-Aldrich

Nikkomycin Z from Streptomyces tendae

≥90% (HPLC)

Synonym(s):

Neopolyoxin C, Nikkomycin Z

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N5O10
CAS Number:
Molecular Weight:
495.44
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

H2O: soluble 5 mg/mL

antibiotic activity spectrum

fungi

Mode of action

cell wall synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

C[C@@H]([C@H](N)C(=O)N[C@H]([C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=O)NC2=O)C(O)=O)[C@H](O)c3ccc(O)cn3

InChI

1S/C20H25N5O10/c1-7(13(28)9-3-2-8(26)6-22-9)11(21)17(31)24-12(19(32)33)16-14(29)15(30)18(35-16)25-5-4-10(27)23-20(25)34/h2-7,11-16,18,26,28-30H,21H2,1H3,(H,24,31)(H,32,33)(H,23,27,34)/t7-,11-,12+,13-,14-,15+,16+,18+/m0/s1

InChI key

WWJFFVUVFNBJTN-JKEIIPFCSA-N

General description

Chemical structure: peptidyl nucleoside

Application

Nikkomycin Z is used as a selective competitive inhibitor of chitin synthetase 3 in studies on fungal cell wall development. It is a potential treatment for human Encephalitozoon hellem, a microsporidian species[1] and is used to study the changes of chitin and β--glucan under Nikkomycin Z exposure[2].

Biochem/physiol Actions

Nikkomycin Z inhibits chitin synthase from converting UDP-GlcNAc into cell wall chitin due to its structural resemblance to UDP-N-acetylglucosamine[3].

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L Popolo et al.
Journal of bacteriology, 179(2), 463-469 (1997-01-01)
The GGP1/GAS1 gene codes for a glycosylphosphatidylinositol-anchored plasma membrane glycoprotein of Saccharomyces cerevisiae. The ggp1delta mutant shows morphogenetic defects which suggest changes in the cell wall matrix. In this work, we have investigated cell wall glucan levels and the increase
T Chapman et al.
Antimicrobial agents and chemotherapy, 36(9), 1909-1914 (1992-09-01)
An N-acetyl-D-[14C]glucosamine radiolabel incorporation assay has been used to monitor chitin biosynthesis in whole cells of Candida albicans both in vitro and in vivo in two different mouse infection models, one using the peritoneal cavity as a chamber in which
Matthew M Draelos et al.
Nature chemical biology, 17(2), 213-221 (2020-12-02)
Kinases are annotated in many nucleoside biosynthetic gene clusters but generally are considered responsible only for self-resistance. Here, we report an unexpected 2'-phosphorylation of nucleoside biosynthetic intermediates in the nikkomycin and polyoxin pathways. This phosphorylation is a unique cryptic modification
E Bigliardi et al.
Antimicrobial agents and chemotherapy, 44(11), 3012-3016 (2000-10-19)
Since 1985 microsporidia have been recognized as a cause of emerging infections in humans, mainly in immunocompromised human immunodeficiency virus-positive subjects. As chitin is a basic component of the microsporidian infective stage, the spore, we evaluated in vitro the susceptibility
H Decker et al.
Journal of general microbiology, 137(8), 1805-1813 (1991-08-01)
The structure-activity relationships of different nikkomycins were studied to evaluate the structural requirements for a potent chitin synthase inhibitor. We investigated the transport of the nikkomycins via the peptide transport system of the yeast Yarrowia lipolytica and determined the kinetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service