Skip to Content
Merck
All Photos(1)

Key Documents

N7885

Sigma-Aldrich

Neuraminidase from Vibrio cholerae

Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)

Synonym(s):

Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Quality Level

type

Type III

form

buffered aqueous solution

specific activity

1-5 units/mg protein (Lowry, using NAN-lactose)

mol wt

83 kDa

application(s)

diagnostic assay manufacturing

foreign activity

Protease and NAN-aldolase, present

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Neuraminidase enzymes are glycoside hydrolase enzymes that catalyze hydrolysis of terminal sialic acid residues. The most well-known are the viral nearamidases, which promote influenza virus release.

Application

Neuraminidase from Vibrio cholerae has been used in a study to assess its role in the binding and uptake of cholera toxin by susceptible cells. It has also been used in a study to investigate the preparation of a sodium substrate and its use in a fluorometric assay of neuraminidase.
Neurminidase is used as a cell-surface probe for glycoconjugate distribution and in substrate specificity studies.

Biochem/physiol Actions

Vibrio cholerae neuraminidase has been shown to promote cholera toxin infection by binding the toxin and aiding in its uptake by cells.

Quality

Preservative free.

Unit Definition

One unit will liberate 1.0 μmole of N-acetylneuraminic acid per min at pH 5.0 at 37 °C using NAN-lactose or bovine submaxillary mucin, unless otherwise specified. Prices based on units using NAN-lactose as substrate.

Physical form

Aqueous solution, pH 5.5, containing 0.15 M NaCl and 4 mM CaCl2.

Preparation Note

Chromatographically purified.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Rota et al.
Organic & biomolecular chemistry, 10(14), 2885-2894 (2012-03-08)
A simple protocol for the synthesis of N-perfluoroacylated and N-acylated glycals of neuraminic acid, with a secondary cyclic amine (morpholine or piperidine) at the 4α position, has been set-up, starting from peracetylated N-acetylneuraminic acid methyl ester that undergoes, sequentially to
Jai Woong Seo et al.
Nature communications, 11(1), 2102-2102 (2020-05-02)
Adeno-associated viruses (AAVs) are typically single-stranded deoxyribonucleic acid (ssDNA) encapsulated within 25-nm protein capsids. Recently, tissue-specific AAV capsids (e.g. PHP.eB) have been shown to enhance brain delivery in rodents via the LY6A receptor on brain endothelial cells. Here, we create
P C Reading et al.
Journal of virology, 74(11), 5190-5197 (2000-05-09)
Influenza viruses A/PR/8/34 (PR8; H1N1), A/Aichi/68 X-31 (HKx31; H3N2), and A/Beijing/89 X-109 (BJx109; H3N2) show marked differences in their ability to infect murine macrophages, including resident alveolar and peritoneal macrophages as well as the macrophage-derived cell line J774. The hierarchy
Fluorometric assay of neuraminidase with a sodium (4-methylumbelliferyl-alpha-D-N-acetylneuraminate) substrate.
M Potier et al.
Analytical biochemistry, 94(2), 287-296 (1979-04-15)
J E Galen et al.
Infection and immunity, 60(2), 406-415 (1992-02-01)
Vibrio cholerae neuraminidase (NANase) is hypothesized to act synergistically with cholera toxin (CT) and increase the severity of a secretory response by increasing the binding and penetration of CT to enterocytes. To test this hypothesis, the NANase gene (nanH) from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service