Skip to Content
Merck
All Photos(1)

Key Documents

N7009

Sigma-Aldrich

2-Nitro-5-thiocyanatobenzoic acid

≥98% (HPLC)

Synonym(s):

2-Nitro-5-thiocyanobenzoic acid, NTCB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(SCN)CO2H
CAS Number:
Molecular Weight:
224.19
Beilstein:
2124001
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

2-Nitro-5-thiocyanatobenzoic acid, powder

Assay

≥98% (HPLC)

Quality Level

form

powder

color

yellow

mp

156-157 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)c1cc(SC#N)ccc1[N+]([O-])=O

InChI

1S/C8H4N2O4S/c9-4-15-5-1-2-7(10(13)14)6(3-5)8(11)12/h1-3H,(H,11,12)

InChI key

NQUNIMFHIWQQGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Site-Specific Conversion of Cysteine in a Protein to Dehydroalanine Using 2-Nitro-5-thiocyanatobenzoic Acid: This research demonstrates the use of 2-Nitro-5-thiocyanatobenzoic acid in protein engineering to modify cysteine residues, which is critical for developing therapeutic proteins and understanding protein structure-function relationships (Qiao et al., 2021).

  • Scrapie prion protein structural constraints obtained by limited proteolysis and mass spectrometry: Highlights the utility of 2-Nitro-5-thiocyanatobenzoic acid in the study of prion proteins, providing insights into protein misfolding diseases which can lead to novel therapeutic approaches (Sajnani et al., 2008).

  • Localization of a substrate binding domain of the human reduced folate carrier to transmembrane domain 11 by radioaffinity labeling and cysteine-substituted accessibility methods: Utilizes 2-Nitro-5-thiocyanatobenzoic acid in biochemical assays to understand transporter proteins, essential for drug delivery and targeting (Hou et al., 2005).

  • A novel procedure for the preparation of biologically active recombinant peptides using a cyanylation reaction: Describes a method using 2-Nitro-5-thiocyanatobenzoic acid for peptide synthesis, beneficial for the development of peptide-based pharmaceuticals (Koyama et al., 1994).

Biochem/physiol Actions

2-nitro-5-thiocyanatobenzoic acid (NTCB) is commonly used to cyanylate and cleave proteins at cysteine residues.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Daniel et al.
The Journal of biological chemistry, 272(43), 26934-26939 (1997-10-27)
Escherichia coli dihydroorotase contains six cysteines/subunit, which are potential ligands of structural and catalytic zinc metals at protein sites of the enzyme. Specific thiol reagents modify, in nondenaturing conditions only, two of these cysteines; these two residues are thought to
Maya Belghazi et al.
Molecular and cellular endocrinology, 247(1-2), 175-182 (2006-02-07)
Luteinizing hormone (LH) like all other glycoprotein hormones is composed of two dissimilar subunits, alpha and beta, that are non-covalently associated. The heterodimer is stabilized by a region of the beta-subunit called the "seatbelt" because it wraps around the alpha-subunit
Z Xie et al.
Journal of neurochemistry, 93(4), 1038-1046 (2005-04-29)
Apolipoprotein J (apoJ; also known as clusterin and sulfated glycoprotein (SGP)-2) is associated with senile plaques in degenerating regions of Alzheimer's disease brains, where activated microglia are also prominent. We show a functional link between apoJ and activated microglia by
Hsin-Yao Tang et al.
Analytical biochemistry, 334(1), 48-61 (2004-10-07)
The reagent 2-nitro-5-thiocyanatobenzoic acid (NTCB) is commonly used to cyanylate and cleave proteins at cysteine residues, but this two-step reaction requires lengthy incubations and produces highly incomplete cleavages. In previous reports, incomplete cleavage was attributed to a competing beta-elimination reaction
N Tanaka et al.
Biochemistry, 40(20), 5914-5920 (2001-05-16)
We investigated the pressure effect on the conformation of beta-amylase by monitoring the chemical reaction of the unpaired cysteine. Sweet potato beta-amylase is composed of four identical subunits, each of which contains six cysteine residues. These residues are inert to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service