Skip to Content
Merck
All Photos(4)

Key Documents

N3376

Sigma-Aldrich

Nicotinamide

≥98% (HPLC), powder, PARP inhibitor

Synonym(s):

Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein:
383619
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Nicotinamide, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

color

white

mp

128-131 °C (lit.)

application(s)

cell analysis

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Used as a cofactor for certain enzymes

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage and Stability

This product can be stored tightly closed in room temperature and in dry conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian W Johnson et al.
Cell reports, 28(6), 1538-1550 (2019-08-08)
Ras GTPases are mutated at codons 12, 13, and 61, with different frequencies in KRas, HRas, and NRas and in a cancer-specific manner. The G13D mutant appears in 25% of KRas-driven colorectal cancers, while observed only rarely in HRas or
Robert J Motzer et al.
The Lancet. Oncology, 14(6), 552-562 (2013-04-20)
In a phase 3 trial comparing the efficacy and safety of axitinib versus sorafenib as second-line treatment for metastatic renal cell carcinoma, patients given axitinib had a longer progression-free survival (PFS). Here, we report overall survival and updated efficacy, quality
Jordi Bruix et al.
Gut, 63(5), 844-855 (2014-02-18)
Hepatocellular carcinoma (HCC) is one of the leading causes of cancer-related death and is currently the main event leading to death in patients with cirrhosis. Evolving information suggests that the metabolic syndrome with non-alcoholic liver disease may be an important
Ann-Lii Cheng et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(32), 4067-4075 (2013-10-02)
Open-label, phase III trial evaluating whether sunitinib was superior or equivalent to sorafenib in hepatocellular cancer. Patients were stratified and randomly assigned to receive sunitinib 37.5 mg once per day or sorafenib 400 mg twice per day. Primary end point
Xiao-bin Lv et al.
Scientific reports, 5, 17940-17940 (2015-12-15)
SUN2, a key component of LINC (linker of nucleoskeleton and cytoskeleton) complex located at the inner nuclear membrane, plays unknown role in lung cancer. We found that SUN2 expression was decreased in lung cancer tissue compared with paired normal tissues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service