Skip to Content
Merck
All Photos(1)

Documents

N3288

Sigma-Aldrich

(+)-Norfenfluramine hydrochloride

≥98% (HPLC), powder

Synonym(s):

JP 92, NSC 43036, S-(+)-α-Methyl-3-(trifluoromethyl)benzeneethanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12F3N · HCl
CAS Number:
Molecular Weight:
239.67
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

color

white

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cl.C[C@H](N)Cc1cccc(c1)C(F)(F)F

InChI

1S/C10H12F3N.ClH/c1-7(14)5-8-3-2-4-9(6-8)10(11,12)13;/h2-4,6-7H,5,14H2,1H3;1H/t7-;/m0./s1

InChI key

PIDLOFBRTWNFAR-FJXQXJEOSA-N

Biochem/physiol Actions

(+)-Norfenfluramine is the major hepatic metabolite of (+)-fenfluramine and is primarily responsible for the anorexic effect, as well as side effects; (+)-norfenfluramine causes vasoconstriction and a blood pressure increase in rats with normal blood; acts primarily on the cytoplasmic pool of serotonin; more potent than fenfluramine at inducing dopamine release.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service