Skip to Content
Merck
All Photos(3)

Key Documents

N3019

Sigma-Aldrich

2-Naphthoxyacetic acid

suitable for plant cell culture, BioReagent, crystalline

Synonym(s):

BNOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH2CO2H
CAS Number:
Molecular Weight:
202.21
Beilstein:
1074148
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

form

crystalline

technique(s)

cell culture | plant: suitable

color

white to gray

mp

151-154 °C (lit.)

application(s)

agriculture

SMILES string

OC(=O)COc1ccc2ccccc2c1

InChI

1S/C12H10O3/c13-12(14)8-15-11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2,(H,13,14)

InChI key

RZCJYMOBWVJQGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Naphthoxyacetic acid is a plant growth hormone having structure related to auxin and is majorly used to regulate growth of tomatoes, apple and grapes.

Application

β-Naphthoxyacetic acid (BNOA) is a phytohormone used as a plant growth regulator on tomatoes and strawberries.
2-Naphthoxyacetic acid has been used as a component for embryo cell suspension maintenance medium. It has been used as a medium component for inducing somatic embryogenesis in grapevine.

Biochem/physiol Actions

2-Naphthoxyacetic acid promotes seed germination and early flowering in tomatoes. 2-naphthoxyacetic acid effectively improves fruit size and color. However, it is also regarded slightly hazardous and may contribute to mutagenesis.

Preparation Note

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sufang Fan et al.
Journal of separation science, 34(6), 640-650 (2011-02-12)
In the experiment, a high-performance liquid chromatography and electrospray ionization-tandem mass spectrometry with selected reaction monitoring was used to simultaneously determine various classes of phytohormones, including indole-3-acetic acid, α-naphthaleneacetic acid, 2-chlorobenzoic acid, 4-chlorobenzoic acid, indole-3-butyric acid, gibberellic acid, 2,4-dichlorophenoxyacetic acid
Synthesis and controlled release properties of beta-naphthoxyacetic acid intercalated Mg-Al layered double hydroxides nanohybrids
Li S, et al.
Arabian Journal of Chemistry (2015)
Somatic embryogenesis from stigmas and styles of grapevine
Carimi F, et al.
In Vitro Cellular & Developmental Biology. Plant, 41(3), 249-252 (2005)
Plant Cell culture Protocols, 77-86 (2006)
The development and comparison of a fluorescence and a phosphorescence optosensors for determining the plant growth regulator 2-naphthoxyacetic acid
Casado-Terrones S, et al.
Sensors and Actuators B, Chemical, 107(2), 929-935 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service