Skip to Content
Merck
All Photos(4)

Key Documents

N3013

Sigma-Aldrich

Nile Red

Powder

Synonym(s):

Nile Blue A Oxazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18N2O2
CAS Number:
Molecular Weight:
318.37
Beilstein:
279110
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Nile Red, Technical grade

grade

technical grade

form

powder

composition

Carbon: 70.0-77.3%

Nitrogen: 7.5-8.8%

technique(s)

titration: suitable

color

faint green to brown

mp

203-205 °C (lit.)

solubility

methanol: 1 mg/mL

λmax

553 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCN(CC)c1ccc2N=C3C(Oc2c1)=CC(=O)c4ccccc34

InChI

1S/C20H18N2O2/c1-3-22(4-2)13-9-10-16-18(11-13)24-19-12-17(23)14-7-5-6-8-15(14)20(19)21-16/h5-12H,3-4H2,1-2H3

InChI key

VOFUROIFQGPCGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nile Red is a lipophilic fluorescent dye that is commonly used in various scientific applications, particularly in lipid research. It has the ability to stain and visualize lipid droplets in cells and tissues.

Application

Nile red is widely employed in studies related to lipid metabolism, lipid droplet dynamics, lipid accumulation in cells or tissues, and lipid-based drug delivery systems.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-Ying Chen et al.
Plant physiology, 180(2), 813-826 (2019-03-23)
Mitochondrial fission occurs frequently in plant cells, but its biological significance is poorly understood because mutants specifically impaired in mitochondrial fission do not show obvious defects in vegetative growth. Here, we revealed that the production of viable pollen was reduced
Glycine N-methyltransferase deficiency affects Niemann-Pick type C2 protein stability and regulates hepatic cholesterol homeostasis.
Liao YJ
Molecular Medicine, 18, 412-412 (2012)
Zebrafish obesogenic test: a tool for screening molecules that target adiposity.
Tingaud-Sequeira A
Journal of Lipid Research, 52, 1765-1765 (2011)
D L Sackett et al.
Analytical biochemistry, 167(2), 228-234 (1987-12-01)
Nile red is an uncharged hydrophobic molecule whose fluorescence is strongly influenced by the polarity of its environment. It interacts with many, but not all, native proteins, including beta-lactoglobulin, kappa-casein, and albumin, with a wide range of spectral changes for
Autophagy gene disruption reveals a non-vacuolar cell death pathway in Dictyostelium.
Kosta A
The Journal of Biological Chemistry, 279, 48404-48404 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service