MX633S100
Mix-n-Stain™ CF™ 633 Antibody Labeling Kit (50-100μg)
Sign Into View Organizational & Contract Pricing
All Photos(8)
About This Item
Recommended Products
form
liquid
Quality Level
storage condition
protect from light
fluorescence
λex 630 nm; λem 650 nm
storage temp.
−20°C
Related Categories
General description
This labeling kit is for labeling 50-100 μg antibody with the far red fluorescent dye CF633. Mix-n-Stain CF dye antibody labeling kits are a revolutionary antibody labeling technology that permits one to label antibodies with our superior CF dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. CF633 is a novel rhodamine-based far-red dye with excellent fluorescence quantum yield and exceptional photostability. The dye can be efficiently excited by the 633 or 635 nm laser with emission maximum at 650 nm. Both the excitation and emission wavelengths of CF633 are shorter than those of the cyanine-based dye Cy®5 or Alexa Fluor™ 647. Despite the spectral difference, CF633 is still brighter than Alexa Fluor™647 and other similar cyanine dyes when used in flow cytometry applications on popular flow cytometers. However, on other instruments where the detection windows are more optimized for Cy®5-like dyes, the full advantage of CF633 may not be evident. Ensure that instrument settings are optimized for use with CF633 to obtain the most benefit from the dye. The most important advantage of CF633 is its exceptional photostability, which is unmatched by any of the cyaninebased far-red dyes, including Cy®5, Alexa Fluor™ 647 and DyLight® 649.
Application
The Mix-n-Stain CF dye antibody labeling kit can be used to label primary antibody for ELISA, western blot, immunohistochemistry, flow cytometry, immunofluorescence or immunocytochemistry applications.
Features and Benefits
Mix-n-Stain CF dye antibody labeling kits dramatically simplify the process of preparing fluorescently labeled antibodies, particularly primary antibodies. Simply mix your antibody with a CF dye of your choice in the buffer provided, a step that takes less than 30 seconds of hands-on time. After waiting for another 30 minutes, without a separation step, you will have a fluorescent antibody conjugate that is as good as the best pre-labeled fluorescent antibody you can buy from some of the leading suppliers. There is no need to calculate how much dye you should use; just mix your antibody with the entire amount of dye provided and you will always produce optimal labeling. Moreover, the labeling reaction can tolerate the presence of common stabilizers, such as sodium azide, Tris, glycerol and low level of BSA.
Components
The kit consists of 1 vial each of Mix-n-Stain Dye, Mix-n-Stain Reaction Buffer (10x), Mix-n-Stain Storage Buffer, and an ultrafiltration vial.
Analysis Note
Direct replacement for: Alexa Fluor™ 633, DY-630, DyLight® 633
Legal Information
This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.
Cy is a registered trademark of Cytiva
DyLight is a registered trademark of Pierce Biotechnology, Inc.
Mix-n-Stain is a trademark of Biotium, Inc.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Frontiers in immunology, 13, 828319-828319 (2022-03-12)
TIGIT (T-cell immunoglobulin and ITIM domain) has emerged as a promising target in cancer immunotherapy. It is an immune "checkpoint" inhibitor primarily expressed on activated T cells, NK cells and Tregs. Engagement of TIGIT to its ligands PVR and PVR-L2
EBioMedicine, 60, 102996-102996 (2020-09-20)
Pertuzumab is currently used in combination with trastuzumab as the first-line treatment for HER2-positive metastatic breast cancer. However, pertuzumab was originally developed independently from trastuzumab and was later incidentally found to have synergistic efficacy when combined with trastuzumab, it remains
Neuron, 109(12), 1963-1978 (2021-05-26)
Our daily life depends on muscle contraction, a process that is controlled by the neuromuscular junction (NMJ). However, the mechanisms of NMJ assembly remain unclear. Here we show that Rapsn, a protein critical for NMJ formation, undergoes liquid-liquid phase separation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service