Skip to Content
Merck

MTCC2002

MTC Bio SureSeal S Microcentrifuge Tube

capacity 2.0 mL, sterile, clear, pack of 500 ea

Synonym(s):

centrifuge tube, plastic tube, MTC Bio C2002, sample collection tube, sample receptacle, sample tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41121703
Capacity:
2.0 mL
Material:
clear , conical bottom , polypropylene
Sterility:
sterile
Packaging:
pack of 500 ea
Parameter:
28,000 × g speed
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , conical bottom , polypropylene

sterility

sterile

packaging

pack of 500 ea

manufacturer/tradename

MTC Bio C2002

parameter

28,000 × g speed

capacity

2.0 mL

General description

Designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation, and general sample/reagent storage. SureSeal(TM) S tubes are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28,000×g) and the usable temperature range (-90°C to +121°C). The full-sized tube cap (with frosted marking surface), combined with the unique ring and groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps.
Microtube w/ cap, 2.0ml, clear, sterile, w/ self-standing bag & Stop-Pops, 500/pk

Features and Benefits

  • Sterile comes standard – by electron beam radiation
  • Self-Standing bag – with gusseted bottom
  • Stop-Pops supplied – 10 cap locks in every bag

Legal Information

MTC is a trademark of MTC Bio
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Stop-Pops is a trademark of MTC Bio
SureSeal S is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library