Skip to Content
Merck
All Photos(1)

Key Documents

MSST0052

Sigma-Aldrich

SILuLite AKT2, RAC-beta serine/threonine-protein kinase human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

Protein kinase Akt-2, Protein kinase B beta (PKB beta), RAC-PK-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12
Pricing and availability is not currently available.

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... AKT2(208)

Related Categories

General description

SILuLite AKT2 is a recombinant human protein expressed in human 293 cells. It consists of 502 amino acids, with a calculated molecular mass of 58.2 kDa (including N-terminal polyhistidine and flag tags). SILu AKT2 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Sequence

MDYKDDDDKGHHHHHHHHGGQMNEVSVIKEGWLHKRGEYIKTWRPRYFLLKSDGSFIGYKERPEAPDQTLPPLNNFSVAECQLMKTERPRPNTFVIRCLQWTTVIERTFHVDSPDEREEWMRAIQMVANSLKQRAPGEDPMDYKCGSPSDSSTTEEMEVAVSKARAKVTMNDFDYLKLLGKGTFGKVILVREKATGRYYAMKILRKEVIIAKDEVAHTVTESRVLQNTRHPFLTALKYAFQTHDRLCFVMEYANGGELFFHLSRERVFTEERARFYGAEIVSALEYLHSRDVVYRDIKLENLMLDKDGHIKITDFGLCKEGISDGATMKTFCGTPEYLAPEVLEDNDYGRAVDWWGLGVVMYEMMCGRLPFYNQDHERLFELILMEEIRFPRTLSPEAKSLLAGLLKKDPKQRLGGGPSDAKEVMEHRFFLSINWQDVVQKKLLPPFKPQVTSEVDTRYFDDEFTAQSITITPPDRYDSLGLLELDQRTHFPQFSYSASIRE

Physical form

Supplied as a liquid containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service