MSMLS07
MSMLS™ Plate 7 (Lipophilic)
Supplied by IROA Technologies
Synonym(s):
MSMLS, Mass Spectrometry Metabolite Libarary of Standards, Metabolic Library, Metabolite Library
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.28
Recommended Products
form
solid
packaging
pkg of 1 plates
manufacturer/tradename
IROA Technologies
plate size
96 wells
shipped in
dry ice
storage temp.
−20°C
General description
MSMLS Plate 7 is a collection of high quality small biochemical molecules that span a broad range of primary metabolism. These are high purity (>95%) compounds supplied in an economical, ready-to-use format. This library of standards is most commonly used for metabolite qualification and quantification for MS and NMR, functional cellular assays, and phenotypic screening. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound, and Substance ID.The library is intended to be used for mass spectrometry metabolomics applications and provides a broad representation of primary metabolism.
Application
Metabolite qualification and quantification for MS, NMR, etc.; Functional cellular assays; Phenotypic screening. Suitable for manual and automated work flows.
Features and Benefits
To view a complete list of compounds in the library for recent lots, download the plate maps below:
The complete plate map with name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMBD ID, PubChem Compound, and Substance ID are provided with the software download.
Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds:
The complete plate map with name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMBD ID, PubChem Compound, and Substance ID are provided with the software download.
Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds:
- Carboxylic acids, amino acids
- Biogenic amines, polyamines
- Nucleotides, coenzymes and vitamins
- Mono- and disaccharides
- Fatty acids, lipids, steroids and hormones
Preparation Note
MSMLS Plate 7 compounds are conveniently provided at 5 μg per well, enough for multiple injections. Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds by row to allow multiple mixtures to be analyzed per injection. For more information on MSMLS07, solubilization and preparation notes please refer to the product information sheet.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Legal Information
MSMLS is a trademark of IROA Technologies LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Charles M Nichols et al.
Analytical chemistry, 90(24), 14484-14492 (2018-11-20)
In this work, we established a collision cross section (CCS) library of primary metabolites based on analytical standards in the Mass Spectrometry Metabolite Library of Standards (MSMLS) using a commercially available ion mobility-mass spectrometer (IM-MS). From the 554 unique compounds
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service