Skip to Content
Merck
All Photos(1)

Key Documents

MSKT0001

Sigma-Aldrich

SILuMAb K1 Pharmacokinetic Kit

Synonym(s):

PK Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.41

suitability

suitable for mass spectrometry

storage temp.

2-8°C

General description

All critical reagents necessary to perform the assay are provided in the kit. The plate-based format avoids the high cost and potential LC column blockage associated with bead-based enrichment formats. Sample preparation with the SILuMAb PK kit can be performed in under 5 hours and consists of coating wells with high-specificity capture antibody, followed by immunoaffinity enrichment and rapid, in-plate trypsin digestion. The resulting digest is ready for injection, with no SPE or other cleanup required.

Application

The SILuMAb K1 Pharmacokinetic (PK) kit enables a robust, high-throughput assay for quantification of human IgG1 antibody in animal sera by LC-MS/MS.

Storage and Stability

Store the kit at 2-8°C. The kit is stable for two years refrigerated.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service