Skip to Content
Merck
All Photos(2)

Documents

MAK147

Sigma-Aldrich

Mitochondrion Membrane Potential Kit

sufficient for 500 fluorometric tests (orange fluorescence, microplate readers)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 500 fluorometric tests (orange fluorescence, microplate readers)

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Mitochondria generate a potential across their membranes due to the activities of enzymes of the electron transport chain. During apoptosis, collapse of the mitochondrial membrane potential (MMP) coincides with the opening of the mitochondrial permeability transition pores, leading to the release of cytochrome c into the cytosol, which in turn triggers other downstream events in the apoptotic cascade.

Application

Mitochondrion Membrane Potential Kit has been used to estimate the mitochondrion membrane potential in chinese hamster ovary (CHO) cells. It has been used in the permeabilization of the mitochondrial membrane test (MMP assay) and also used to detect the loss of mitochondrial membrane permeabilization.

Suitability

This kit is suitable for the detection of mitochondrial membrane potential (MMP) in mammalian cells and for screening apoptosis inhibitors and activators using microplate readers.

Principle

Mitochondrion Membrane Potential Kit is optimized for the detection of the loss of the MMP in cells. The cationic hydrophobic mitochondrial potential dye accumulates in normal mitochondria, most likely due to the mitochondrial potential, resulting in an increase in fluorescence (λex = 540/λem = 590 nm). In apoptotic cells, MMP collapse results in decreased fluorescence. This kit can be used for monitoring apoptosis and for screening apoptosis inhibitors and activators.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ngoc An Le et al.
Journal of medicinal chemistry, 64(10), 6792-6801 (2021-05-15)
Photodynamic therapy (PDT) is used to treat various cancerous diseases. Recently, we have demonstrated that platinated pyridyl-substituted porphyrins are potent agents for PDT with very high phototoxicity (IC50 down to 17 nM) and excellent phototoxic indices of higher than 5800
A zombie LIF gene in elephants is upregulated by TP53 to induce apoptosis in response to DNA damage
Vazquez J M, et al.
Cell Reports, 24(7), 1765-1776 (2018)
George K Latsis et al.
International journal of molecular sciences, 19(7) (2018-07-18)
Two known tin-based polymers of formula {[R₃Sn(CH₃COO)]n} where R = n-Bu⁻ (1) and R = Ph⁻ (2),were evaluated for their in vitro biological properties. The compounds were characterized via their physical properties and FT-IR, 119Sn Mössbauer, and ¹H NMR spectroscopic
Juan Manuel Vazquez et al.
Cell reports, 24(7), 1765-1776 (2018-08-16)
Large-bodied organisms have more cells that can potentially turn cancerous than small-bodied organisms, imposing an increased risk of developing cancer. This expectation predicts a positive correlation between body size and cancer risk; however, there is no correlation between body size
New metalo-therapeutics of NSAIDs against human breast cancer cells.
Banti C N, et al.
European Journal of Medicinal Chemistry, 143, 1687-1701 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service