Skip to Content
Merck
All Photos(1)

Key Documents

MAK120

Sigma-Aldrich

Urease Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Urea Amidohydrolase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

application(s)

environmental

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases

storage temp.

2-8°C

General description

Urease is an enzyme that catalyzes the hydrolysis of urea into carbon dioxide and ammonia. Urease is found in bacteria, yeast, and certain higher plant species. Urease activity is found in environmental and fecal samples.

Application

Urease Activity Assay Kit has been used to measure urease activity.

Features and Benefits

Compatible with high-throughput handling systems. Can be adapted for use with cuvettes.

Suitability

Suitable for the detection of Urease activity in biological and environmental samples.

Principle

In this assay, urease catalyzes the hydrolysis of urea resulting in the production of ammonia. The ammonia is determined by the Berthelot method resulting in colorimetric product measure at 670 nm, proportionate to the urease activity present in the sample. One unit of urease is the amount of enzyme that catalyzes the formation of 1.0 μmole ammonia per minute at pH 7.0.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelsea M Heveran et al.
Scientific reports, 9(1), 14721-14721 (2019-10-13)
We demonstrate for the first time that the morphology and nanomechanical properties of calcium carbonate (CaCO3) can be tailored by modulating the precipitation kinetics of ureolytic microorganisms through genetic engineering. Many engineering applications employ microorganisms to produce CaCO3. However, control
Ureases I. Functional, catalytic and kinetic properties: A review
Barbara K.
Journal of Molecular Catalysis. B, Enzymatic, 59(1-3), 9-21 (2009)
Bhawna Uprety et al.
Pharmaceutics, 14(1) (2022-01-22)
Tumour cells maintain a local hypoxic and acidic microenvironment which plays a crucial role in cancer progression and drug resistance. Urease is a metallohydrolases that catalyses the hydrolysis of urea into ammonia and carbon dioxide, causing an abrupt increase of
Satya P G
Hydroxamic Acids: A Unique Family of Chemicals with Multiple Biological Activities (2013)
James M Wiebler et al.
Proceedings. Biological sciences, 285(1878) (2018-05-04)
Gut bacteria that produce urease, the enzyme hydrolysing urea, contribute to nitrogen balance in diverse vertebrates, although the presence of this system of urea-nitrogen recycling in Amphibia is as yet unknown. Our studies of the wood frog (Rana sylvatica), a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service