Skip to Content
Merck
All Photos(1)

Key Documents

M9697

Sigma-Aldrich

MST1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp686A2068, KRS2, STK4, YSK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

952-1288 nmol/min·mg

mol wt

~83 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STK4(6789)

Biochem/physiol Actions

MST1 belongs to a family of proteins that share similarity with a budding yeast serine/threonine kinase, sterile-20 (Ste20). Endogenous full-length MST1 is activated by a variety of stressful stimuli, accompanied by the secondary appearance of a 36 kDa Thr183-phosphorylated, caspase-cleaved catalytic fragment. Recombinant MST1 undergoes a robust autoactivation in vitro, mediated by an intramolecular autophosphorylation on the activation loop of an MST dimer. MST1 can initiate apoptosis when transiently overexpressed in mammalian cells. Interference with the ability of endogenous MST1 to associate with the putative tumor suppressor proteins Nore1/RASSF can inhibit Ras-induced apoptosis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Avruch et al.
Methods in enzymology, 407, 290-310 (2006-06-08)
The six human Nore1/RASSF genes encode a family of putative tumor suppressor proteins, each expressed as multiple mRNA splice variants. The predominant isoforms of these noncatalytic polypeptides are characterized by the presence in their carboxyterminal segments of a Ras-Association (RA)
P M de Souza et al.
Biochemical Society transactions, 32(Pt3), 485-488 (2004-05-26)
Mammalian Sterile20-like kinase 1 (Mst1) is a ubiquitously expressed serine/threonine kinase which represents a member of the rapidly expanding family of enzymes related to the yeast Sterile20 kinase. Although the physiological function of Mst1 and its role in intracellular signalling
Yavuz Kulaberoglu et al.
Nature communications, 8(1), 695-695 (2017-09-28)
The Hippo tumor suppressor pathway is essential for development and tissue growth control, encompassing a core cassette consisting of the Hippo (MST1/2), Warts (LATS1/2), and Tricornered (NDR1/2) kinases together with MOB1 as an important signaling adaptor. However, it remains unclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service