Skip to Content
Merck
All Photos(1)

Key Documents

M8693

Sigma-Aldrich

Melanotan II acetate salt

≥95% (HPLC)

Synonym(s):

Ac-[Nle4Asp5D-Phe7Lys10]α-MSH-(4-10)-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H69N15O9 · xC2H4O2
CAS Number:
Molecular Weight:
1024.18 (free base basis)
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

color

white

solubility

H2O: soluble 5 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[nH]1cncc1C[C@@H]2NC(=O)[C@H](CC(=O)NCCCC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](NC2=O)Cc5ccccc5)CCCN=C(N)N)Cc3c4c([nH]c3)cccc4)C(=O)N)NC(=O)[C@@H](NC(=O)C)CCCC

InChI

1S/C50H69N15O9/c1-3-4-16-36(59-29(2)66)44(69)65-41-25-42(67)55-20-11-10-18-35(43(51)68)60-47(72)39(23-31-26-57-34-17-9-8-15-33(31)34)63-45(70)37(19-12-21-56-50(52)53)61-46(71)38(22-30-13-6-5-7-14-30)62-48(73)40(64-49(41)74)24-32-27-54-28-58-32/h5-9,13-15,17,26-28,35-41,57H,3-4,10-12,16,18-25H2,1-2H3,(H2,51,68)(H,54,58)(H,55,67)(H,59,66)(H,60,72)(H,61,71)(H,62,73)(H,63,70)(H,64,74)(H,65,69)(H4,52,53,56)/t35-,36-,37-,38+,39-,40-,41-/m0/s1

InChI key

JDKLPDJLXHXHNV-MFVUMRCOSA-N

Gene Information

Related Categories

Amino Acid Sequence

Ac-Nle-Asp-His-Phe-Arg-Trp-Lys-NH2

General description

Melanotan II induces melanogenesis and thus increases skin pigmentation. It is an analog of α-melanocyte stimulating hormone.

Application

Melanotan II acetate salt has been used to study its inhibitory effects on feeding and energy expenditure in rats.

Biochem/physiol Actions

Melanotan II is a MC3-R/MC4-R melanocortin agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristian Thestrup-Pedersen et al.
Ugeskrift for laeger, 173(13), 975-975 (2011-04-02)
A 25-year-old man had self-injected more than 150 doses of melanotan to increase his skin pigmentation, which had increased significantly. At the same time, his nevi had become darker and new nevi and lentigines developed; they also occurred on his
Rosacea 101: Includes the Rosacea Diet null
Miia Kilpeläinen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 77(1), 20-25 (2010-10-23)
Peptide molecules can improve the treatment of a number of pathological conditions, but due to their physicochemical properties, their delivery is very challenging. The study aim was to determine whether nanostructured porous silicon could sustain the release and prolong the
Yi Zhang et al.
Peptides, 31(4), 639-643 (2009-12-26)
Central melanocortins (MC) evoke potent but transient anorectic responses with tachyphylaxis developing within days. We hypothesized that intermittent therapy using the MC analog, melanotan II (MTII), would minimize the tachyphylaxis and enhance the long-term efficacy of MTII treatment. F344/BN rats
Y B Shrestha et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 299(1), R140-R149 (2010-04-23)
Norepinephrine (NE) released from the sympathetic nerves innervating white adipose tissue (WAT) is the principal initiator of lipolysis in mammals. Central WAT sympathetic outflow neurons express melanocortin 4-receptor (MC4-R) mRNA. Single central injection of melanotan II (MTII; MC3/4-R agonist) nonuniformly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service