Skip to Content
Merck
All Photos(3)

Key Documents

M6546

Sigma-Aldrich

(±)-2-(p-Methoxyphenoxy)propionic acid

solid, ≥98%

Synonym(s):

PMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%

biological source

synthetic (organic)

Assay

≥98%

form

solid

SMILES string

COc1ccc(OC(C)C(O)=O)cc1

InChI

1S/C10H12O4/c1-7(10(11)12)14-9-5-3-8(13-2)4-6-9/h3-7H,1-2H3,(H,11,12)

InChI key

MIEKOFWWHVOKQX-UHFFFAOYSA-N

Application

(±)-2-(p-Methoxyphenoxy)propionic acid may be used as a sweet taste receptor antagonist to study its effect on allulose-induced glucagon-like peptide 1 (GLP-1) secretion rats. It may also be used to study the response of the secretin tumor cell line (STC-1) to sweetener in the presence and absence of (±)-2-(p-methoxyphenoxy)propionic acid or lactisole.

Biochem/physiol Actions

As a taste modulator, the sodium salt inhibits the perception of sweetness in humans, but not in rats.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sclafani et al.
Physiology & behavior, 61(1), 25-29 (1997-01-01)
Cypha, propionic acid, 2-(4-methoxyphenol) salt, is a commercially available sweet taste inhibitor used in food products. The present study examined whether or not Cypha blocked the sweet taste response of rats. This was accomplished by measuring the consummatory response of
David E Lanfear et al.
JACC. Heart failure, 5(11), 823-832 (2017-11-04)
This study sought to derive and validate plasma metabolite associations with survival in heart failure (HF) patients. Profiling of plasma metabolites to predict the course of HF appears promising, but validation and incremental value of these profiles are less established. Patients
Kensuke Ninomiya et al.
The EMBO journal, 39(3), e102729-e102729 (2019-11-30)
A number of long noncoding RNAs (lncRNAs) are induced in response to specific stresses to construct membrane-less nuclear bodies; however, their function remains poorly understood. Here, we report the role of nuclear stress bodies (nSBs) formed on highly repetitive satellite
Almudena Aguilera-Diaz et al.
PloS one, 15(1), e0227986-e0227986 (2020-01-25)
The diagnosis of myeloid neoplasms (MN) has significantly evolved through the last few decades. Next Generation Sequencing (NGS) is gradually becoming an essential tool to help clinicians with disease management. To this end, most specialized genetic laboratories have implemented NGS
Zhihao Zhang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 1350-1358 (2016-11-03)
Dendrobium officinale Kimura et Migo (Dendrobium catenatum Lindley), a prized traditional Chinese Medicine, has been used in China and Southeast Asian countries for centuries. The present study was aimed to investigate the effects and the possible mechanisms of the Dendrobium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service