Skip to Content
Merck
All Photos(3)

Key Documents

M6145

Sigma-Aldrich

1-Thioglycerol

liquid, BioReagent, suitable for cell culture, ≥97% (titration)

Synonym(s):

α-Monothioglycerol, α-Thioglycerol, 3-Mercapto-1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
108.16
Beilstein:
1732046
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

product line

BioReagent

Assay

≥97% (titration)

form

liquid

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

refractive index

n20/D 1.527 (lit.)

bp

118 °C/5 mmHg (lit.)

solubility

water: miscible 0.1 M

density

1.25 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(O)CS

InChI

1S/C3H8O2S/c4-1-3(5)2-6/h3-6H,1-2H2

InChI key

PJUIMOJAAPLTRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture studies of embryonic cortical and hippocampal neurons, mouse bone marrow mast cell lines, and human Β cell lines have utilized 1-thioglycerol as a component of the culture medium to stimulate proliferation.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morgan Oatley et al.
Nature communications, 11(1), 586-586 (2020-01-31)
The endothelial to haematopoietic transition (EHT) is the process whereby haemogenic endothelium differentiates into haematopoietic stem and progenitor cells (HSPCs). The intermediary steps of this process are unclear, in particular the identity of endothelial cells that give rise to HSPCs
L Tang et al.
Journal of biomedical materials research, 41(2), 333-340 (1998-06-25)
Biomaterial-mediated complement activation repeatedly has been invoked as a trigger of phagocyte reactions and inflammation. However, a direct correlation between complement activation and inflammatory responses to biomaterial surfaces has yet to be established. Using an animal implantation model and gold
Suji Lee et al.
Stem cells (Dayton, Ohio), 36(10), 1552-1566 (2018-07-14)
Although autologous induced pluripotent stem cells (iPSCs) can potentially be useful for treating patients without immune rejection, in reality it will be extremely expensive and labor-intensive to make iPSCs to realize personalized medicine. An alternative approach is to make use
Yohsuke Hanaoka et al.
PloS one, 9(4), e94930-e94930 (2014-04-17)
Tissue inhibitors of metalloproteinases (TIMPs) regulate matrix metalloproteinase activity and maintain extracellular matrix homeostasis. Although TIMP-3 has multiple functions (e.g., apoptosis, inhibition of VEGF binding to VEGF receptor, and inhibition of TNFα converting enzyme), its roles in thermogenesis and metabolism
Ceri-Anne E Suurmond et al.
Advanced healthcare materials, 8(24), e1901379-e1901379 (2019-11-21)
The liver has a complex and unique microenvironment with multiple cell-cell interactions and internal vascular networks. Although nonalcoholic fatty liver disease (NAFLD) is the most common chronic liver disease with multiple phases, no proper model could fully recapitulate the in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service