Skip to Content
Merck
All Photos(1)

Key Documents

M4937

Sigma-Aldrich

Anti-Melanocortin-3 Receptor antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MC3R

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40 kDa

species reactivity

rat, mouse (predicted)

technique(s)

microarray: suitable
western blot: 1:2,000 using rat brain homogenate extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Mc3r(17201)
rat ... Mc3r(29310)

General description

Melanocortins are regulatory peptides derived from post-translational processing of the larger pro-opiomelanocortin (POMC) precursor. Melanocortin-3 Receptor (MC3-R) is a 360 amino acid containing transmembrane protein that belongs to G-protein coupled receptor family and is expressed mainly in adult central nervous system at high levels in a restricted number of neurons of the hypothalamus and the limbic system.

Immunogen

A synthetic peptide corresponding to the N-terminal region of rat melanocortin receptor 3 (amino acids 15-33) coupled to keyhole limpet hemocyanin (KLH). This sequence is highly conserved in mouse MC3-R (89% identity).

Application

Anti-Melanocortin-3 Receptor antibody produced in rabbit has been used in western blotting[1] and immunocytochemistry.

Biochem/physiol Actions

Melanocortin-3 (MC3) receptor has a crucial role in facilitating the effects of the melanocortin system on energy homeostasis. The MC3 receptor has been proposed to mediate the actions of MSH, in particular those of α-MSH and γ-MSH peptides. MC3-R is thought to regulate fat stores by a metabolic pathway. MC3-R knockout mice (MC3-R-/-) have increased fat mass, but not high body mass index, are hyperleptinaemic and develop mild hyperinsulinaemia. In contrast to MC3-R, MC4-R primarily controls food intake, suggesting that both MC3 and MC4 receptors may play a complementary role in weight control.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mc3 and Mc4 receptors: complementary role in weight control
Raffin S M L, et al.
European Journal of Endocrinology, 144(1), 207-208 (2019)
Inflamed phenotype of the mesenteric microcirculation of melanocortin type 3 receptor-null mice after ischemia-reperfusion
Leoni G, et al.
Faseb Journal, 22(12), 4228-4238 (2008)
Paul M Holloway et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(9), 1936-1944 (2015-06-27)
Neutrophil recruitment is a key process in the pathogenesis of stroke, and may provide a valuable therapeutic target. Targeting the melanocortin (MC) receptors has previously shown to inhibit leukocyte recruitment in peripheral inflammation, however, it is not known whether treatments
Magdalena K Kaneva et al.
British journal of pharmacology, 167(1), 67-79 (2012-04-05)
Melanocortin MC(1) and MC(3 ) receptors, mediate the anti-inflammatory effects of melanocortin peptides. Targeting these receptors could therefore lead to development of novel anti-inflammatory therapeutic agents. We investigated the expression of MC(1) and MC(3) receptors on chondrocytes
A S Chen et al.
Nature genetics, 26(1), 97-102 (2000-09-06)
Genetic and pharmacological studies have defined a role for the melanocortin-4 receptor (Mc4r) in the regulation of energy homeostasis. The physiological function of Mc3r, a melanocortin receptor expressed at high levels in the hypothalamus, has remained unknown. We evaluated the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service