Skip to Content
Merck
All Photos(1)

Key Documents

M3315

Sigma-Aldrich

MJ33 lithium salt

powder, ≥90% (NMR)

Synonym(s):

1-Hexadecyl-3-(trifluoroethyl)-sn-glycero-2-phosphomethanol lithium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43F3O6PLi
CAS Number:
Molecular Weight:
498.48
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥90% (NMR)

form

powder

color

white to beige

solubility

H2O: ≥5 mg/mL (warmed at 60 °C)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCOCC(COCC(F)(F)F)OP(OC)([O-])=O.[Li+]

InChI

1S/C22H44F3O6P.Li/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-29-18-21(31-32(26,27)28-2)19-30-20-22(23,24)25;/h21H,3-20H2,1-2H3,(H,26,27);/q;+1/p-1

InChI key

GDLMLQJISATCEL-UHFFFAOYSA-M

Application

MJ33 lithium salt has been used as a peroxiredoxin VI (Prdx6) inhibitor:
  • to study its specific Prdx6 peroxidase activity in vitro and in breast cancer cell line (MCF-7) cell lysates
  • to study its effects on a mouse model of acute lung injury associated with exposure to hyperoxia
  • to study its potential use as a therapeutic agent

Biochem/physiol Actions

MJ33 acts as a non-toxic and potent inhibitor of agonist-induced activation of nicotinamide adenine dinucleotide phosphate (NADPH) oxidase type 2 (NOX2). It also prevents lung injuries associated with lung inflammation in mice. MJ33 is a fluorinated lipid analog, which blocks the enzyme by mimicking the transition state of the substrate.
Novel, active site directed, specific, competitive and reversible inhibitor of phospholipase A2 (PLA2).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Manevich et al.
Free radical biology & medicine, 54, 62-70 (2012-11-13)
The dual-functioning antioxidant enzyme peroxiredoxin VI (Prdx6) detoxifies lipid peroxides particularly in biological membranes, and its peroxidase function is activated by glutathione S-transferase Pi (GSTP). The GSTP gene is polymorphic in humans, with the wild-type GSTP1-1A (Ile105, Ala114) and three
Inhibition of the phospholipase A2 activity of peroxiredoxin 6 prevents lung damage with exposure to hyperoxia
Benipal B, et al.
Redox Biology, 4, 321-327 (2015)
Bavneet Benipal et al.
Redox biology, 4, 321-327 (2015-02-01)
Lung injury associated with hyperoxia reflects in part the secondary effects of pulmonary inflammation and the associated production of reactive oxygen species due to activation of NADPH oxidase, type 2 (NOX2). Activation of NOX2 requires the phospholipase A2 (PLA2) activity
Allelic variants of glutathione S-transferase P1-1 differentially mediate the peroxidase function of peroxiredoxin VI and alter membrane lipid peroxidation
Manevich Y, et al.
Free radical biology & medicine, 54, 62-70 (2013)
Fisher, A.B. et al.
American Journal of Physiology. Cell Physiology, 280, 748-748 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service