Skip to Content
Merck
All Photos(1)

Key Documents

M0894

Sigma-Aldrich

Minimum Essential Medium Eagle

Alpha Modification, with L-glutamine and sodium pyruvate, without ribonucleosides, deoxyribonucleosides and sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

stable glutamine: no
Hanks’ salts (2% CO2): no
sodium pyruvate: yes
NaHCO3: no
phenol red: yes
glucose: yes
L-glutamine: yes
Earle’s salts (5% CO2): yes
HEPES: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins.

Quantity

Formulated to contain 10.1 grams of powder per liter of medium.

Reconstitution

Supplement with 2.2 g/L sodium bicarbonate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason W Ashley et al.
The Journal of biological chemistry, 291(44), 23268-23281 (2016-10-30)
Macrophages are important in innate and adaptive immunity. Macrophage participation in inflammation or tissue repair is directed by various extracellular signals and mediated by multiple intracellular pathways. Activation of group VIA phospholipase A2 (iPLA2β) causes accumulation of arachidonic acid, lysophospholipids
Alexander J Nelson et al.
Journal of lipid research, 61(2), 143-158 (2019-12-11)
Phospholipases A2 (PLA2s) catalyze hydrolysis of the sn-2 substituent from glycerophospholipids to yield a free fatty acid (i.e., arachidonic acid), which can be metabolized to pro- or anti-inflammatory eicosanoids. Macrophages modulate inflammatory responses and are affected by Ca2+-independent phospholipase A2
Tatsuki Kawasaki et al.
Development, growth & differentiation, 58(6), 530-545 (2016-06-01)
The periodontal ligament (PDL) is a connective tissue that attaches the tooth cementum to the alveolar bone and is derived from dental follicle cells (DFCs). The DFCs form fibroblasts, osteoblasts, cementoblasts, and PDL stem cells (PDLSCs). We previously reported homeobox
Kateřina Valentová et al.
International journal of molecular sciences, 19(8) (2018-08-12)
Silymarin, an extract from milk thistle (Silybum marianum) fruits, is consumed in various food supplements. The metabolism of silymarin flavonolignans in mammals is complex, the exact structure of their metabolites still remains partly unclear and standards are not commercially available.
Jun Zhang et al.
Therapeutic advances in chronic disease, 12, 2040622321995685-2040622321995685 (2021-08-31)
Previous studies reported that melatonin exerts its effect on mesenchymal stem cell (MSC) survival and differentiation into osteogenic and adipogenic lineage. In the current study we aimed to explore the effect of melatonin on osteoporosis and relevant mechanisms. Real-time qualitative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service