Skip to Content
Merck
All Photos(1)

Documents

M0450

Sigma-Aldrich

Minimum Essential Medium Eagle

Alpha Modification, with nucleosides, ʟ-glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Minimum Essential Medium Eagle, Alpha Modification, With ribonucleosides, L-glutamine, deoxyribonucleosides and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

glucose: 1.0 g/L
L-glutamine: 0.292 g/L
phenol red: 0.011 g/L
NaHCO3: 2.2 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media. This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins. See formulation for details. Often used in cytogenetic analysis when supplemented with FBS.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of chemically defined cell culture media-replacing fetal bovine serum in mammalian in vitro methods
Van der VJ, et al.
Toxicology in vitro, 24(4), 1053-1063 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service