LSMLS04
LSMLS™ Plate 4 (Water Soluble)
Supplied by IROA Technologies
Synonym(s):
LSMLS, Large Scale Metabolite Libarary of Standards, Metabolic Library, Metabolite Library
About This Item
Recommended Products
form
solid
packaging
pkg of 1 plates
manufacturer/tradename
IROA Technologies
plate size
96 wells
shipped in
dry ice
storage temp.
−20°C
General description
Application
Features and Benefits
The complete plate map with name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMBD ID, PubChem Compound, and Substance ID are provided with the software download.
Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds:
- Carboxylic acids, amino acids
- Biogenic amines, polyamines
- Nucleotides, coenzymes and vitamins
- Mono- and disaccharides
- Fatty acids, lipids, steroids and hormones
Preparation Note
Other Notes
Legal Information
Signal Word
Danger
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 1A - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 - STOT RE 1 Inhalation - STOT RE 2 - STOT SE 3
Target Organs
Blood, Lungs, Respiratory system
Storage Class Code
3 - Flammable liquids
Flash Point(C)
30 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service