Skip to Content
Merck
All Photos(1)

Key Documents

L9536

Sigma-Aldrich

Anti-Leptin Receptor antibody produced in goat

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunohistochemistry (frozen sections): suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Lepr(16847)

Immunogen

recombinant mouse leptin receptor expressed in NS0 cells.

Biochem/physiol Actions

Leptin is a hormone secreted by adipocytes and regulates energy homeostasis, inflammation and neuroendocrine processes, synaptic formation and memory condensation. The physiological effects of leptin are mediated by leptin receptors, ObR and LepR. Leptin is the key regulator of obesity and acts directly on neuronal networks to regulate feeding, metabolism, gastric mobility and energy homeostasis. The pathways that mediate the effect of leptin are JAK/STAT, PI3K and JNK.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liwei Jiang et al.
Frontiers in immunology, 12, 730438-730438 (2022-02-04)
Lymph node (LN)-resident stromal cells play an essential role in the proper functioning of LNs. The stromal compartment of the LN undergoes significant compensatory changes to produce a milieu amenable for regulation of the immune response. We have identified a
Sami M Khan et al.
Yonsei medical journal, 53(4), 671-679 (2012-06-06)
Leptin, a peptide hormone secreted by adipocytes in proportion of the amount of energy stored in fat, plays a central role in regulating human energy homeostasis. In addition, leptin plays a significant permissive role in the physiological regulation of several
Laura Marroquí et al.
Journal of molecular endocrinology, 49(1), R9-17 (2012-03-27)
Leptin plays an important role in the control of food intake, energy expenditure, metabolism, and body weight. This hormone also has a key function in the regulation of glucose homeostasis. Although leptin acts through central and peripheral mechanisms to modulate
A G Smith et al.
Developmental biology, 121(1), 1-9 (1987-05-01)
Many pluripotent embryonal carcinoma (EC) cell lines and all embryonic stem (ES) cell lines have hitherto been maintained in the undifferentiated state only by culture on feeder layers of mitomycin C-treated embryonic fibroblasts. We now demonstrate that medium conditioned by
Weihong Pan et al.
Annals of the New York Academy of Sciences, 1264, 64-71 (2012-04-26)
Leptin, an adipocyte-derived cytokine, crosses the blood-brain barrier to act on many regions of the central nervous system (CNS). It participates in the regulation of energy balance, inflammatory processes, immune regulation, synaptic formation, memory condensation, and neurotrophic activities. This review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service