Skip to Content
Merck
All Photos(1)

Key Documents

L6920

Sigma-Aldrich

LFM-A13

≥98% (HPLC), powder

Synonym(s):

α-Cyano-β-hydroxy-β-methyl-N-(2,5-dibromophenyl)propenamide, (2Z)-2-Cyano-N-(2,5-dibromophenyl)-3-hydroxy-2-butenamide; a-Cyano-b-hydroxy-b-methyl-N-(2,5-dibromophenyl)propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8Br2N2O2
CAS Number:
Molecular Weight:
360.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥19 mg/mL

storage temp.

2-8°C

SMILES string

C\C(O)=C(/C#N)C(=O)Nc1cc(Br)ccc1Br

InChI

1S/C11H8Br2N2O2/c1-6(16)8(5-14)11(17)15-10-4-7(12)2-3-9(10)13/h2-4,16H,1H3,(H,15,17)/b8-6-

InChI key

UVSVTDVJQAJIFG-VURMDHGXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

LFM-A13 is an inhibitor of the nonreceptor tyrosine kinase Bruton′s tyrosine kinase (Btk), a new molecular target for treatment of B-cell lymphoma and autoimmune disorders. LFMA13 inhibited recombinant BTK with an IC50 value of 2.5microM with 100-fold lower inhibitory activity against a number of other protein kinases including JAK1, JAK2, EGFR, IRK, IKK, and CDK1, 2 and 3, as well as several other related kinases

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Wang et al.
Oncotarget, 8(21), 34099-34110 (2017-04-19)
Tec kinase, a prototypical member of the Tec tyrosine kinases family, was shown to mainly govern lymphocyte proliferation. In the present study, we investigated the role of Tec kinase in acute inflammatory response in lipopolysaccharide (LPS) challenge. First, we demonstrate
Yuanyuan Li et al.
Oncotarget, 8(30), 49238-49252 (2017-05-18)
Bone marrow X-linked kinase (BMX, also known as Etk) has been reported to be involved in cell proliferation, differentiation, apoptosis, migration and invasion in several types of tumors, but its role in cervical carcinoma remains poorly understood. In this study
Wei Zhang et al.
BioMed research international, 2020, 3152043-3152043 (2020-07-21)
Tec kinase is an important mediator in inflammatory immune response that enhances the activity of neutrophils and macrophages. However, information on its function in lipopolysaccharide- (LPS-) induced acute kidney injury (AKI) is limited. This study is aimed at determining whether
Sandra Olsson et al.
Journal of inflammation (London, England), 3, 8-8 (2006-05-06)
Yeast and bacteria elicit arachidonate release in macrophages, leading to the formation of leukotrienes and prostaglandins, important mediators of inflammation. Receptors recognising various microbes have been identified, but the signalling pathways are not entirely understood. Cytosolic phospholipase A2 is a
Karoline Gäbler et al.
Journal of cellular and molecular medicine, 17(2), 265-276 (2013-01-11)
The Janus kinase 2 mutant V617F occurs with high frequency in myeloproliferative neoplasms. Further mutations affecting the Janus kinase family have been discovered mostly in leukaemias and in myeloproliferative neoplasms. Owing to their involvement in neoplasia, inflammatory diseases and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service