Skip to Content
Merck
All Photos(2)

Key Documents

L5037

Sigma-Aldrich

Leptin from rat

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

OB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
biological source:
Escherichia coli
form:
lyophilized powder

biological source

Escherichia coli

Quality Level

recombinant

expressed in E. coli

sterility

non-sterile

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

≤0.8 ng/mL ED50

mol wt

16 kDa

impurities

≤1.0 EU/μg endotoxin

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

rat ... Lep(25608)

Application

Leptin from rat has been used:
  • to check the effect of intranasal nerve growth factor (NGF) on NGF activity in the spinal cord of Sprague-Dawley rats
  • for validation of labelling for leptin-bearing cells of Sprague-Dawley rats
  • to study the effect of leptin on STAT3 (signal transducer and activator of transcription 3) phosphorylation in neural population of Sprague-Dawley rats
  • to study leptin response towards chemoreflex in nucleus of the solitary tract
  • in rats, to check the effect of association between leptin and neuronal nitric oxide pathway on penicillin-induced epileptiform activity

Biochem/physiol Actions

Hormone produced primarily in adipocytes; primary site of action appears to be on neurons in the hypothalamus that are involved in regulating energy balance, appetite, and body weight.
In rodents with uncontrolled diabetes mellitus, leptin is crucial for normalizing hyperglycemia. In the digestive tract, it also participates in the absorption of dietary molecules via controlling transporters activity.
Leptin is a hormone produced primarily in adipocytes, although leptin mRNA has also been identified in placenta and fetal tissues, gastric tissue and liver. Its primary site of action appears to be on neurons in the hypothalamus that are involved in regulating energy balance, appetite, and body weight. Leptin increases the production of nitric oxide in endothelial cells and stimulates angiogenesis in vitro and in vivo.
Human and mouse leptin share ~84% sequence identity.

Physical form

Lyophilized from a 0.2 μm filtered solution with 4.5 mM NaHCO3.

Analysis Note

The biological activity is measured by its ability to induce proliferation of leptin-dependent human OB-R transfected murine BAF3 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luigi Aloe et al.
Neural regeneration research, 9(10), 1025-1030 (2014-09-11)
The purpose of this work was to investigate whether, by intranasal administration, the nerve growth factor bypasses the blood-brain barrier and turns over the spinal cord neurons and if such therapeutic approach could be of value in the treatment of
Ali Aslan et al.
Brain research, 1321, 117-124 (2010-02-02)
The aim of the present study was to investigate the role of NO as a mediator of leptin action at the penicillin-induced epileptiform activity in rat. Thirty minutes after penicillin injection, leptin, at a dose of 1 microg, significantly increased
Iris A McDuffie et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 52(2), 263-273 (2004-01-20)
Leptin, the ob protein, regulates food intake and satiety and can be found in the anterior pituitary. Leptin antigens and mRNA were studied in the anterior pituitary (AP) cells of male and female rats to learn more about its regulation.
John Ciriello et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 303(7), R727-R736 (2012-08-24)
Circulating levels of leptin are elevated in individuals suffering from chronic intermittent hypoxia (CIH). Systemic and central administration of leptin elicits increases in sympathetic nervous activity (SNA), arterial pressure (AP), and heart rate (HR), and it attenuates the baroreceptor reflex
Carmen Fanjul et al.
Journal of physiology and biochemistry, 71(2), 311-317 (2015-05-04)
Leptin is secreted into the digestive tract and contributes to the absorption of dietary molecules by regulating transporters activity. Here, we studied the effect of luminal leptin on the intestinal transport of L-glutamate, an important component of human diet. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service