Skip to Content
Merck
All Photos(2)

Key Documents

L4900

Sigma-Aldrich

Lonidamine

mitochondrial hexokinase inhibitor

Synonym(s):

1-(2,4-Dichlorobenzyl)-1H-indazole-3-carboxylic acid, Diclondazolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10Cl2N2O2
CAS Number:
Molecular Weight:
321.16
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

chloroform: methanol (1:1): 9.80-10.20 mg/mL, clear, colorless to light yellow

SMILES string

OC(=O)c1nn(Cc2ccc(Cl)cc2Cl)c3ccccc13

InChI

1S/C15H10Cl2N2O2/c16-10-6-5-9(12(17)7-10)8-19-13-4-2-1-3-11(13)14(18-19)15(20)21/h1-7H,8H2,(H,20,21)

InChI key

WDRYRZXSPDWGEB-UHFFFAOYSA-N

Application

Lonidamine has been used:
  • as an inhibitor of glycolysis to study its effects on the epithelial-mesenchymal transition (EMT) mechanism in cancer stem cells (CSC)
  • as mitochondrial permeability transition (MPT)- inducing drug to study its effects on temozolomide-resistant glioblastoma cell line
  • as mitochondrial hexokinase inhibitor to measure reactive oxygen species (ROS) in human acute myeloid leukemia cells

Biochem/physiol Actions

Inhibits the energy metabolism of neoplastic cells by interfering with hexokinase and disrupting uncoupler-stimulated mitochondrial electron transport; damages cell and mitochondrial membranes.
Lonidamine (LND) is an indazole derivative and is an anti-spermatogenic agent. It exhibits anti-tumor activity by sensitizing tumors to photodynamic, chemo-, radiotherapy, and hyperthermia. LND also interferes with energy metabolism by blocking glycolysis, adenosine diphosphate (ADP-), and uncoupler-stimulated respiration on various nicotinamide adenine dinucleotide (NAD-) and flavin adenine dinucleotide (FAD)-linked substrates, hexokinase activity, and oxygen uptake in Ehrlich ascites tumor cells. It blocks lactate export and pyruvate uptake into the mitochondria by inhibiting proton-linked monocarboxylate transporter (MCT) and mitochondrial pyruvate carrier (MPC), respectively.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Calviño et al.
Biochemical pharmacology, 82(11), 1619-1629 (2011-09-06)
Lonidamine is a safe, clinically useful anti-tumor drug, but its efficacy is generally low when used in monotherapy. We here demonstrate that lonidamine efficaciously cooperates with the anti-leukemic agent arsenic trioxide (ATO, Trisenox) to induce apoptosis in HL-60 and other
Morgane Le Bras et al.
Cancer research, 66(18), 9143-9152 (2006-09-20)
Mitochondrial membrane permeabilization (MMP) is a rate-limiting step of apoptosis, including in anticancer chemotherapy. Adenine nucleotide translocase (ANT) mediates the exchange of ADP and ATP on the inner mitochondrial membrane in healthy cells. In addition, ANT can cooperate with Bax
Marilisa P Dimmito et al.
Biomolecules, 9(9) (2019-09-19)
The endocannabinoid system represents an integrated neuronal network involved in the control of several organisms' functions, such as feeding behavior. A series of hybrids of 5-(4-chlorophenyl)-1-(2,4-dichloro-phenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide (mimonabant), a well-known inverse agonist of the type-1 cannabinoid receptor (CB1), once used as
Yolanda Sánchez et al.
The Journal of pharmacology and experimental therapeutics, 335(1), 114-123 (2010-07-08)
Arsenic trioxide (ATO, Trisenox) is an important antileukemic drug, but its efficacy is frequently low when used as a single agent. Here, we demonstrate that the apoptotic action of ATO is greatly increased when combined with subcytotoxic curcumin concentrations in
Lara Milane et al.
Nanomedicine : nanotechnology, biology, and medicine, 7(4), 435-444 (2011-01-12)
The aim of this study was to assess the biodistribution and pharmacokinetics of epidermal growth factor receptor (EGFR)-targeted polymer-blend nanoparticles loaded with the anticancer drugs lonidamine and paclitaxel. Plasma, tumor, and tissue distribution profiles were quantified in an orthotopic animal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service