Skip to Content
Merck
All Photos(1)

Key Documents

J4252

Sigma-Aldrich

JWH-015

≥98% (HPLC), powder

Synonym(s):

(2-Methyl-1-propyl-1H-indol-3-yl)-1-naphthalenylmethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21NO
CAS Number:
Molecular Weight:
327.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

off-white

solubility

DMSO: >20 mg/mL (with heating)

storage temp.

−20°C

SMILES string

CCCn1c(C)c(C(=O)c2cccc3ccccc23)c4ccccc14

InChI

1S/C23H21NO/c1-3-15-24-16(2)22(20-12-6-7-14-21(20)24)23(25)19-13-8-10-17-9-4-5-11-18(17)19/h4-14H,3,15H2,1-2H3

InChI key

LJSBBBWQTLXQEN-UHFFFAOYSA-N

Gene Information

human ... CNR2(1269)
rat ... Cnr1(25248)

Application

JWH-015 has been used to study its effect on tumor-evoked pain behavior. It has also been used to study its protective effects on rat brain mitochondria upon paraquat exposure.

Biochem/physiol Actions

JWH-015 is a selective CB2 cannabinoid receptor agonist. JWH-015 is structurally classified as naphthoylindoles.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intrathecal Administration of the Cannabinoid 2 Receptor Agonist JWH015 Can Attenuate Cancer Pain and Decrease mRNA Expression of the 2B Subunit of N-Methyl-D-Aspartic Acid
XiaoPing G, et al.
Journal of Cell Science, jcs-100925 (2012)
Joseph E Henriquez et al.
Toxicology and applied pharmacology, 369, 82-89 (2019-02-27)
Plasmacytoid dendritic cells (pDC) compose 0.2-0.5% of circulating leukocytes but play a significant role in mounting host immune responses. Elevated and chronic activation of pDC are implicated in autoimmune disease like systemic lupus erythematosus and rheumatoid arthritis. Δ9-tetrahydrocannabinol (THC) is
Protective effects of the synthetic cannabinoids CP55, 940 and JWH-015 on rat brain mitochondria upon paraquat exposure
Velez-Pardo C, et al.
Neurochemical Research, 35(9), 1323-1332 (2010)
G Griffin et al.
European journal of pharmacology, 339(1), 53-61 (1998-02-05)
We have investigated whether there are cannabinoid CB2 receptors that can mediate cannabinoid-induced inhibition of electrically evoked contractions in the mouse vas deferens or guinea-pig myenteric plexus-longitudinal muscle preparation. Our results showed that mouse vas deferens and guinea-pig whole gut
Clinical Toxicology, An Issue of Emergency Medicine Clinics of North America, 4-4 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service