Skip to Content
Merck
All Photos(1)

Key Documents

I5529

Sigma-Aldrich

IgG3, Lambda from human myeloma plasma

purified immunoglobulin, >90% (microfluidic capillary gel electrophoresis)

Synonym(s):

Human IgG3-λ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
application:
citations:
1
Pricing and availability is not currently available.

biological source

human

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

Assay

>90% (microfluidic capillary gel electrophoresis)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Identity of IgG3, lambda light chain is verified by subclass monoclonal antibodies and type specific polyclonal antibodies in immunoelectrphoresis and indirect ELISA assays.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.[1] IgG3 is the has a unique extended hinge region conferring greater flexibility
IgG3, λ from human myeloma plasma is purified by fractionation, ion-exchange and affinity chromatography.
Mammalian immunoglobins contain either lambda or kappa light chains.

Application

IgG3, lambda light chain may be used as an antigen for the production and evaluation of antibodies developed for the indentification and classification of IgG3 subtypes. The purified IgG3, lambda may be used as an immunoglobulin calibrator, reference antigen, blocking agent or coating protein in a variety of immunoassays including ELISA, dot-blot immunobinding, Western immunoblotting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell-binding assays.
The purified IgG3, λ may be used as an immunoglobulin calibrator, reference antigen, blocking agent or coating protein in a variety of immunoassays including ELISA, dot-blot immunobinding, Western immunoblotting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell-binding assays.

Physical form

Solution in 20 mM tris buffered saline, pH 8.0.

Analysis Note

The purity and identity are determined by immunoelectrophoresis, indirect ELISA, and SDS-PAGE.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S Hashira et al.
Pediatrics international : official journal of the Japan Pediatric Society, 42(4), 337-342 (2000-09-15)
Maternal immunoglobulin G (IgG), transferred across the placenta to the fetus during intrauterine life, is an important component of the neonatal immunological defence mechanisms against infection. There is controversy with respect to differences in placental transfer of the different IgG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service