Skip to Content
Merck
All Photos(1)

Key Documents

I3392

Sigma-Aldrich

Anti-Interleukin-1α antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-IL-1α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

indirect ELISA: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il1a(16175)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced by the activation of NF-κ B transcription factor in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signalling through IL-1 receptor type 1. A variety of cells such as macrophages, epithelial cells, fibroblasts, keratinocytes, monocytes and dendritic cells secrete IL-1α. Both IL-1α and IL-1β are important in acquired immunity, inflammation, response to infection and production of matrix metalloproteinases (MMPs). The effects of IL-1α are mediated by activation of NF-κB, JNK and MAPK pathways that activated the genes in response to infection and inflammation. Overproduction of IL-1α has been implicated in inflammatory disorders like rheumatoid arthritis and periodontitis
Anti-interleukin-1α specifically reacts with mouse IL-1α. It does not react with human IL-1α and IL-1β or mouse IL-1β.

Specificity

The antibody will neutralize the biological activity of recombinant mouse IL-1α, but not recombinant human IL-1α, IL-1β or recombinant mouse IL-1β.

Immunogen

recombinant mouse IL-1α.

Application

Anti-interleukin 1α antibody is suitable for neutralization reactions and the recommended ND50 is 0.04-0.2 μg/ml. The antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. It is also suitable for indirect ELISA applications.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline pH 7.4, with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-1 (IL-1) pathway
Weber A et al
Science Signaling, 3, p-p (2010)
Eun-Bum Kang et al.
Apoptosis : an international journal on programmed cell death, 18(11), 1332-1347 (2013-08-03)
Alzheimer's disease (AD) is characterized by the deposition of aggregated amyloid-beta (Aβ), which triggers a cellular stress response called the unfolded protein response (UPR). The UPR signaling pathway is a cellular defense system for dealing with the accumulation of misfolded
S Z Ben-Sasson et al.
Cytokine, 56(1), 122-125 (2011-08-17)
IL-1 strikingly enhances antigen-driven responses of CD4 and CD8 T cells. It is substantially more effective than LPS and when added to a priming regime of antigen plus LPS, it strikingly enhances cell expansion. The effect is mediated by direct
Charles A Dinarello
Current opinion in pharmacology, 4(4), 378-385 (2004-07-15)
All biological agents currently used for reducing TNFalpha activity in disease are neutralization strategies; however, there are several strategies for reducing interleukin (IL)-1 activities: the IL-1 receptor antagonist (IL-1Ra), anti-IL-1beta monoclonal antibodies, the IL-1 Trap, IL-1 receptor type I antibodies
H E Barksby et al.
Clinical and experimental immunology, 149(2), 217-225 (2007-06-26)
Understanding cytokine immunobiology is central to the development of rational therapies for destructive inflammatory diseases such as rheumatoid arthritis (RA) and periodontitis. The classical interleukin-1 (IL-1) family cytokines, IL-1alpha and IL-1beta, as well as IL-18, play key roles in inflammation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service