Skip to Content
Merck
All Photos(1)

Documents

I2513

Sigma-Aldrich

Monoclonal Anti-Human IgG1 antibody produced in mouse

clone 8c/6-39, ascites fluid

Synonym(s):

Monoclonal Anti-Human IgG1 (Fc specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

8c/6-39, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:400

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Human IgG1 (mouse IgG2a isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Human IgG consists of four subclasses (1-4) that can be recognized by antigen differences in their heavy chains. They constitute approximately 65, 25, 6 and 4% of the total IgG, respectively. Each subclass has different biological and physiochemical properties.
The IgG subclass may be preferentially produced in response to different antigens. For instance, antipolysaccharide responses are mainly of the IgG2 subclass, while protein antigens give rise to IgG1 and IgG3 antibodies. Lipopolysaccahrides stimulate an IgG2 response in PBL′s and an IgG1 response in the spleen. Human IgG1 is the predominant subclass of in vivo and in vitro produced anti-tetanus toxoid antibodies. Only IgG1 and IgG3 are capable of adherence to mononuclear phagocytes. Serum IgG subclass deficiencies have been recorded for different patient groups. For example, IgG2 and IgG4 deficiency is associated with IgA deficiency as found in patients of ataxia telangiextasia. Low IgG2 levels were found in patients with SLE and juvenile diabetes melitus. A disproportionate elevation of IgG1 has also been found in the cerebral spinal fluid of patients with multiple sclerosis. Examination of the distribution pattern of IgG subclasses in different types of diseases may provide insight into the immunological processes involved and may assist in the diagnosis of various disorders.
The antibody is specific for the Fc portion of human IgG1 and is non-reactive with other IgG subclasses. This clone, also described as HP-6091, was found to exhibit a valuable profile of reactivity and specificity for human IgG1 by the IUIS/WHO study.

Specificity

Human IgG consists of four subclasses (1-4) that can be recognized.

Application

Monoclonal Anti-Human IgG1 antibody produced in mouse has been used in:
  • direct hemagglutination(HA)
  • hemagglutination inhibition(HAI) assays
  • enzyme linked immunosorbent assay (ELISA)
  • immunofluorometric assay (IFMA)
  • immunofluorescence
  • cell-based assays for the detection of IgG subclasses of acetylcholine receptor (AChR) antibodies
Mouse monoclonal clone clone 8c/6-39 anti-Human IgG1 antibody may be used for the identification of human IgG1 subclass by means of various immunoassays and direct hemagglutination (HA) and hemagglutination inhibition (HAI) assays, enzyme linked immunosorbent assay (ELISA), immunofluorometric assay (IFMA) and detection of cytoplasmic IgG. The antibody may be useful in the analysis of the distribution of IgG subclasses in various diseases.

Biochem/physiol Actions

Human IgG1 is the predominant subclass of in vivo and in vitro produced anti-tetanus toxoid antibodies. Only IgG1 and IgG3 are capable of adherence to mononuclear phagocytes via Fc receptors (FcR). A disproportionate elevation of IgG1 has also been found in the cerebral spinal fluid of patients with multiple sclerosis.

Physical form

The antibody is provided as ascites fluid with 0.1% sodium azide as a preservative.

Storage and Stability

For continuous use, store at 0-5 °C. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chong Yan et al.
Journal of neuroimmunology, 349, 577403-577403 (2020-09-30)
Juvenile-onset myasthenia gravis (JOMG) is a unique clinical subtype in China, featured by a higher prevalence of ocular myasthenia gravis (OMG), higher seronegativity of acetylcholine receptor (AChR) antibodies, and better prognosis than that in adult-onset myasthenia gravis (AOMG). We previously
Determination of IgG Subclasses and Avidity of Antithyroid Peroxidase Antibodies in Patients with Subclinical Hypothyroidism A Comparison with Patients with Overt Hypothyroidism
Silva LM, et al.
Hormone Research in p?diatrics, 59(3), 118-124 (2003)
Wenbo Zhou et al.
Biomedicines, 10(11) (2022-11-12)
Glioblastoma (GBM) is the most aggressive and lethal form of brain tumor. Extracellular vesicles (EVs) released by tumor cells play a critical role in cellular communication in the tumor microenvironment promoting tumor progression and invasion. We hypothesized that GBM EVs
Chong Yan et al.
Pediatric neurology, 98, 74-79 (2019-07-17)
Patients in China with juvenile-onset myasthenia gravis present early, with a high prevalence of purely ocular symptoms, spontaneous remission rates, and low antibody seropositivity. Antibody detection using a cell-based assay has been reported to increase the diagnostic sensitivity in adult-onset
Victor Crivianu-Gaita et al.
Biochemistry and biophysics reports, 2, 23-28 (2015-04-25)
Antibodies have widespread applications in areas ranging from therapeutics to chromatography and protein microarrays. Certain applications require only the fragment antigen-binding (Fab) units of the protein. This study compares the cleavage efficacy of dithiothreitol (DTT), mercaptoethylamine (MEA), and dithiobutylamine (DTBA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service