Skip to Content
Merck
All Photos(1)

Key Documents

I0380

Sigma-Aldrich

D-allo-Isoleucine

≥98% (HPLC)

Synonym(s):

(2R,3S)-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721794
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

D-allo-Isoleucine,

Assay

≥98% (HPLC)

form

powder

optical activity

[α]20/D −38° in 6 M HCl

color

white to off-white

mp

291 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC[C@H](C)[C@@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5+/m0/s1

InChI key

AGPKZVBTJJNPAG-CRCLSJGQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-allo-Isoleucine is used in the synthesis of a conjugate of epi-jasmonic acid and various antimicrobial peptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Koichi Narita et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(42), 11174-11186 (2009-09-18)
The bicyclic depsipeptide histone deacetylase (HDAC) inhibitors spiruchostatins A and B, 5''-epi-spiruchostatin B and FK228 were efficiently synthesized in a convergent and unified manner. The synthetic method involved the following crucial steps: i) a Julia-Kocienski olefination of a 1,3-propanediol-derived sulfone
G Kreil
Ciba Foundation symposium, 186, 77-85 (1994-01-01)
Over the past three decades, numerous peptides have been isolated from amphibian skin secretions. Many of these peptides were shown to be homologous to hormones and neurotransmitters of mammals. In recent years it has been shown that these secretions also
N Ogawa et al.
Amino acids, 42(5), 1955-1966 (2011-05-13)
The TES ether of the C6-hydroxy derivative of naturally occurring epi-jasmonic acid (epi-JA) was designed as epimerization-free equivalent of epi-JA. The TES ether was synthesized from (1R,4S)-4-hydroxycyclopent-2-enyl acetate in 13 steps. The acid part of the ether was activated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service