Skip to Content
Merck
All Photos(5)

Key Documents

I0250

Sigma-Aldrich

Imidazole

≥99% (titration), crystalline

Synonym(s):

1,3-Diaza-2,4-cyclopentadiene, Glyoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.25

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99% (titration)

form

crystalline

impurities

≤0.005% Fluorescence blank (β-NADH equivalence.)

color

white

pH

6.2-7.8 (25 °C)

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

solubility

water: 500 mg/mL, clear, colorless to very faintly yellow

storage temp.

room temp

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Excellent for buffers in the range of pH 6.2-7.8

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Canning et al.
Journal of molecular biology, 426(13), 2457-2470 (2014-04-29)
The discoidin domain receptors (DDRs), DDR1 and DDR2, form a unique subfamily of receptor tyrosine kinases that are activated by the binding of triple-helical collagen. Excessive signaling by DDR1 and DDR2 has been linked to the progression of various human
Pam M Van Ry et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(8), 1285-1297 (2015-06-09)
Duchenne muscular dystrophy (DMD) is a fatal neuromuscular disease caused by mutations in the dystrophin gene, leading to the loss of a critical component of the sarcolemmal dystrophin glycoprotein complex. Galectin-1 is a small 14 kDa protein normally found in skeletal
Ali Tebbi et al.
The Journal of biological chemistry, 290(22), 14077-14090 (2015-04-17)
Ribonucleotide reductase (RnR) is a key enzyme synthesizing deoxyribonucleotides for DNA replication and repair. In mammals, the R1 catalytic subunit forms an active complex with either one of the two small subunits R2 and p53R2. Expression of R2 is S
William A Gaines et al.
Nature communications, 6, 7834-7834 (2015-07-29)
The conserved budding yeast Rad51 paralogues, including Rad55, Rad57, Csm2 and Psy3 are indispensable for homologous recombination (HR)-mediated chromosome damage repair. Rad55 and Rad57 are associated in a heterodimer, while Csm2 and Psy3 form the Shu complex with Shu1 and
Michael D Clark et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(28), E3669-E3678 (2015-07-01)
Acetylation is correlated with chromatin decondensation and transcriptional activation, but its regulation by histone deacetylase (HDAC)-bearing corepressor complexes is poorly understood. Here, we describe the mechanism of assembly of the mammalian Sin3L/Rpd3L complex facilitated by Sds3, a conserved subunit deemed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service