Key Documents
HS14521C
Aluminium weighing boat
D × H 70 mm × 12 mm , large
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
manufacturer/tradename
Heathrow Scientific HS14521C
D × H
70 mm × 12 mm , large
General description
designed for general-purpose weighing, dispensing, storage, food processing, evaporating, or drying.
Boats also make good dust covers for open vessels.
  • All weigh boats are FDA/food-grade compliant
  • Disposable
  • Aluminum, Weighing Boats
Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity. Finger tab handle can be etched for identification prior to weighing.
3 sizes
Packs of 100