HS120845
Rectangular Capsule/Tablet Counting Tray
sterile; electron beam irradiated
Synonym(s):
pill counting tray
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
white polystyrene
sterility
sterile; electron beam irradiated
feature
to hold size 00 to 2 tablets and capsules
packaging
pack of 10 ea
manufacturer/tradename
Heathrow Scientific 120845
L × W × H
10.3 in. × 5.2 in. × 0.5 in.
Related Categories
General description
Intended for single use to prevent cross-contamination for batch sampling or distribution of tablets and capsules.
For Capsule/Tablets size 00 to 2.
Available in both, sterile* and non-sterile.
For Capsule/Tablets size 00 to 2.
Available in both, sterile* and non-sterile.
Features and Benefits
Safety
- Disposable to ensure no chemical cross-contamination for patient safety
- Made from FDA food grade polystyrene in an ISO Class 8 clean room
- Meets quality control and GMP requirements for manufacturing batch sampling
- Durable and shatterproof trays constructed of inert, BSE/TSE free material
- Pour spout in corner for dispensing control
- Ergonomic spatula (length - 6.9 in/17.6 cm
Choose from one of the most recent versions:
Certificates of Analysis (COA)