Skip to Content
Merck
All Photos(4)

Key Documents

HPA054371

Sigma-Aldrich

Anti-IL37 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FIL1, Anti-FIL1(ZETA), Anti-FIL1Z, Anti-IL-1F7, Anti-IL-1H4, Anti-IL1F7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LYCDKDKGQSHPSLQLKKEKLMKLAAQKESARRPFIFYRAQVGSWNMLESAAHPGWFICTSCNCNEPVGVTDKFENRKHIEFSFQPVCKAEMSPSE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL37(27178)

General description

Interleukin 37 (IL-37), also known as IL-1 family member 7 (IL-1F7), is an anti-inflammatory cytokine. It is encoded by the gene mapped to human chromosome 2q12-q14.1. IL-37 is a member of the IL-1 cytokine family. The protein is expressed in blood monocytes, endothelial cells, adipocytes and epithelial cells.

Immunogen

interleukin 37

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-IL37 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Interleukin 37 (IL-37) is implicated in the suppression innate immunity and acquired immune response. IL-37 protects animals from inflammation and autoimmune diseases by inhibiting the production of pro-inflammatory cytokines and by activating macrophage and dendritic cells (DCs). It also exhibits protective action against atherosclerosis. Mutation in the gene is associated with the development of autoimmune thyroid disease (AITD) including Graves′ disease (GD) and Hashimoto′s thyroiditis (HT).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79041

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transgenic overexpression of IL-37 protects against atherosclerosis and strengthens plaque stability
Liu J, et al.
Cellular Physiology and Biochemistry, 45(3), 1034-1050 (2018)
Jiajun Zhou et al.
ImmunoHorizons, 5(10), 830-843 (2021-10-28)
Atopic dermatitis (AD) is a chronic inflammatory skin disease that affects up to one in five children and millions of adults in developed countries. Clinically, AD skin lesions manifest as subacute and/or chronic lichenified eczematous plaques, which are often intensely
Association between interleukin 37 (rs3811047) polymorphism and multiple autoimmune diseases in a Chinese population: A PRISMA-compliant meta-analysis
Lin X, et al.
Medicine, 97(15) (2018)
Epidermal cornification is preceded by the expression of a keratinocyte-specific set of pyroptosis-related genes
Lachner J, et al.
Scientific reports, 7(1), 17446-17446 (2017)
Interleukin-1 as a key factor in the development of inflammatory diseases
Farivar S, et al.
Archives of Pediatric Infectious Diseases, 2(4), 1672-1672 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service