Skip to Content
Merck
All Photos(4)

Key Documents

HPA051535

Sigma-Aldrich

Anti-NELL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ45906, Anti-IDH3GL, Anti-NEL-like 1 (chicken)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

CHCEKTCQVSGLLYRDQDSWVDGDHCRNCTCKSGAVECRRMSCPPLNCSPDSLPVHIAGQCCKVCRPKCIYGGKVLAEGQRILTKSCRECRGGVLVKITEMCPPLNCSEKDHILPENQCCRVCRGHNFCAEGPKCGENSECK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NELL1(4745)

Immunogen

NEL-like 1 (chicken) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86866

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shane A Bobart et al.
Mayo Clinic proceedings, 96(3), 577-591 (2021-03-07)
To describe the clinical and pathological phenotype of membranous nephropathy (MN) associated with M-type-phospholipase-A2-receptor (PLA2R), thrombospondin-type-1-domain-containing-7A (THSD7A), semaphorin 3B (SEMA3B), neural-epidermal-growth-factor-like-1-protein (NELL-1), protocadherin 7 (PCDH7), exostosin 1/exostosin 2 (EXT1/EXT2) and neural cell adhesion molecule 1 (NCAM-1) as target antigens. A
Johannes Münch et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 21(9), 3175-3179 (2021-04-19)
Membranous nephropathy (MN) constitutes a major cause of nephrotic syndrome (NS) in adults. After kidney transplantation (KTx), both recurrent and de novo MN has been reported. In addition to PLA2R and THSD7A, recent identification of neural EGFL-like-1 protein, NELL1, as
Rupali S Avasare et al.
Kidney international reports, 9(5), 1379-1386 (2024-05-06)
Neural epidermal growth factor like 1 membranous nephropathy (NELL1 MN) is associated with various secondary etiologies. However, previous studies on the frequency of these associations and their impact on outcomes are limited. We report a large multiinstitutional series of patients
Dalia Zubidat et al.
Kidney international reports, 8(6), 1213-1219 (2023-06-07)
Membranous nephropathy (MN) is the most common glomerular disease associated with sarcoidosis. The target antigen M-type phospholipase A2 receptor 1 (PLA2R) has been identified in a subset of sarcoidosis-associated MN. The target antigen is not known in the remaining sarcoidosis-associated
Paul Miller et al.
Pediatric nephrology (Berlin, Germany), 37(12), 3127-3137 (2022-03-26)
Membranous nephropathy is an uncommon cause of nephrotic syndrome in pediatrics. We reviewed our kidney biopsy records for patients ≤ 20 years of age with membranous nephropathy without evidence of systemic lupus erythematosus within 6 months of biopsy (January 1995-September

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service