Skip to Content
Merck
All Photos(5)

Key Documents

HPA030751

Sigma-Aldrich

Anti-OGT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Anti-FLJ23071, Anti-HRNT1, Anti-MGC22921, Anti-O-GLCNAC, Anti-O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human, mouse, rat
citations:
8
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LYKIDPSTLQMWANILKRVPNSVLWLLRFPAVGEPNIQQYAQNMGLPQNRIIFSPVAPKEEHVRRGQLADVCLDTPLCNGHTTGMDVLWA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OGT(8473)

General description

OGT (O-linked N-acetylglucosamine transferase) gene is mapped to human chromosome Xq13.1. This gene encodes for nucleo cytosolic and mitochondrial isoforms of the enzyme. OGT is highly conserved among Caenorhabditis elegans, rat, and human genomes, sharing over 65% amino acid identity. The gene includes multiple tandem tetratricopeptide repeat sequences and is modified by O-GlcNA (O-linked N-acetylglucosamine), also by tyrosine phosphorylation.

Immunogen

O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-OGT antibody produced in rabbit has been used in western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

O-GlcNAc (O-linked N-acetylglucosamine) transferase (OGT) catalyzes O-GlcNAcylation that occurs in mitochondria, nuclear, and cytoplasmic proteins. O-GlcNAc transferase is a subunit of nonspecific lethal complex, a transcriptional regulator. O-GlcNAcylation involves the addition of O-GlcNAc to specific serine or threonine residue on many of the cytosolic and nuclear proteins. Disturbance in O-GlcNAcylation is associated with the development of diseases including cancer, diabetes, and Alzheimer disease. OGT is associated with increased mitochondrial respiration and elevated glycolysis. Inhibition in addition or removal of O-GlcNAc leads to a defect in preinitiation complex assembly.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78284

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The O-GlcNAc transferase gene resides on the X chromosome and is essential for embryonic stem cell viability and mouse ontogeny.
Shafi R
Proceedings of the National Academy of Sciences of the USA, 97(11), 5735-5739 (2000)
Suppression of OGT by microRNA24 reduces FOXA1 stability and prevents breast cancer cells invasion.
Liu Y
Biochemical and Biophysical Research Communications, 487(3), 755-762 (2017)
Human RNA Polymerase II Promoter Recruitment in Vitro Is Regulated by O-Linked N-Acetylglucosaminyltransferase (OGT).
Lewis BA
The Journal of Biological Chemistry, 291(27), 14056-14061 (2016)
O-Linked N-acetylglucosamine transferase 1 regulates global histone H4 acetylation via stabilization of the nonspecific lethal protein NSL3.
Wu D
The Journal of Biological Chemistry, 292(24), 10014-10025 (2017)
Mitochondrial O-GlcNAc Transferase (mOGT) Regulates Mitochondrial Structure, Function, and Survival in HeLa Cells.
Sacoman JL
The Journal of Biological Chemistry, 292(11), 4499-4518 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service