Skip to Content
Merck
All Photos(5)

Documents

HPA024372

Sigma-Aldrich

Anti-S100A8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CFAG, Anti-Calgranulin-A, Anti-Calprotectin L1L subunit, Anti-Cystic fibrosis antigen, Anti-Leukocyte L1 complex light chain, Anti-MRP-8, Anti-Migration inhibitory factor-related protein 8, Anti-P8, Anti-Protein S100-A8, Anti-S100 calcium-binding protein A8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

LTELEKALNSIIDVYHKYSLIKGNFHAVYRDDLKKLLETECPQYIRKKGADVWFKELDINTDGAVNFQEFLILVIKM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A8(6279)

General description

The gene S100A8 (S100 calcium-binding protein A8) is mapped to human chromosome 1q21. It is a small calcium-binding protein, which is strongly expressed in neutrophils. In presence of cell stimulation, it is also expressed in monocytes, macrophages, platelets and epithelial and endothelial cells. S100A8 protein is present as a homodimer as well as heterodimer (S100A8/A9). The protein has two EF-hand motifs and one high-affinity Ca2+ binding site. S100A8 is also referred to as myeloid-related protein 8 (MRP8) or calgranulin A.

Immunogen

Protein S100-A8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-S100A8 antibody produced in rabbit has been used in immunohistochemistry and immunofluorescence.

Biochem/physiol Actions

S100A8 (S100 calcium-binding protein A8) is mainly present at the inflammation site or in the serum during acute and chronic inflammation. It is responsible for neutrophil recruitment, attachment and release from the bone marrow. S100A8 is required for sending neutrophils to the site of inflammation in presence of bacterial infection, lipopolysaccharide, and monosodium urate crystals. It is also involved in granulocyte and monocyte attachment to endothelium and also their transport across endothelial cells. The S100A8/A9 heterodimer induces monocyte migration and cytokine secretion. In presence of falciparum malaria, high levels of S100A8 are present in the blood.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70709

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mRNA Expression of S100A8 as a Prognostic Marker for Progression of Non-Muscle-Invasive Bladder Cancer.
Ha YS, et al.
Korean Journal of Urology, 51, 15-20 (2010)
Gabriella Béke et al.
Frontiers in immunology, 9, 424-424 (2018-03-21)
The immunological barrier of the healthy skin is considered to be unified on the whole body surface-however, recent indirect findings have challenged this dogma since microbial and chemical milieu (e.g., sebum, sweat, and pH) exhibit remarkable differences on topographically distinct
Z Dajnoki et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 36(3), 462-471 (2021-11-02)
Hidradenitis suppurativa (HS) is a chronic, inflammatory disease of the apocrine gland-rich (AGR) skin region. The initial steps of disease development are not fully understood, despite intense investigations into immune alterations in lesional HS skin. We aimed to systematically investigate
Secretion of S100A8, S100A9, and S100A12 by Neutrophils Involves Reactive Oxygen Species and Potassium Efflux.
Tardif MR, et al.
Journal of immunology research, 296149-296149 (2015)
Up-regulated S100 calcium binding protein A8 in Plasmodium-infected patients correlates with CD4(+)CD25(+)Foxp3 regulatory T cell generation.
Kim TS, et al.
Malaria Journal, 14, 385-385 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service