Skip to Content
Merck
All Photos(5)

Key Documents

HPA023918

Sigma-Aldrich

Anti-STC1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-STC-1, Anti-Stanniocalcin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

EQNDSVSPRKSRVAAQNSAEVVRCLNSALQVGCGAFACLENSTCDTDGMYDICKSFLYSAAKFDTQGKAFVKESL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STC1(6781)

General description

STC1 (stanniocalcin 1) is a hypocalcemic hormone, which is produced and released by corpuscles of Stannius (CS), an endocrine gland found in bony fish. It is a glycoprotein and in humans, it is widely expressed such as in heart, lung, adrenal gland, liver, kidney, spleen, ovary, prostate, bone and colon.

Immunogen

Stanniocalcin-1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

STC1 (stanniocalcin 1) is responsible for calcium homeostasis in mammals. However, unlike its counterpart in fishes, it functions in a paracrine/autocrine manner in mammals. Studies in mice show that this hormone is primarily involved in carcinogenesis and inflammatory processes in mammals. This gene is significantly up-regulated in human lung, breast, ovary, liver, pancreas, and colon cancer samples. It decreases the size of tumor in HCC (hepatocellular carcinoma) due to its pro-apoptotic and anti-inflammatory actions. Thus, it might have potential therapeutic functions in patients with inflammatory tumors in HCC. Silencing of this gene in ccRCC (clear cell renal cell carcinoma) resulted in prevention of cell proliferation, invasion and metastasis, whereas suppression of this gene resulted in inhibition of EMT (epithelial-mesenchymal transition).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76321

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomonobu Ezure et al.
BioFactors (Oxford, England), 45(6), 944-949 (2019-07-28)
Dermal-epidermal interaction plays a role in many pathophysiological processes, such as tumor invasion and psoriasis, as well as wound healing, and is mediated at least in part by secretory factors. In this study, we investigated the factor(s) involved. We found
Bonnie H Y Yeung et al.
PloS one, 10(10), e0139977-e0139977 (2015-10-16)
Growing evidence has revealed high expression levels of stanniocalcin-1 (STC1) in different types of human cancers. Numerous experimental studies using cancer cell lines demonstrated the involvement of STC1 in inflammatory and apoptotic processes; however the role of STC1 in carcinogenesis
Cherry C T Leung et al.
PloS one, 15(11), e0241932-e0241932 (2020-11-07)
Human stanniocalcin-1 (STC1) is a glycoprotein known to participate in inflammation and tumor progression. However, its role in cancer-macrophage interaction at the tumor environment is not known. In this study, the co-culture of the human metastatic hepatocellular carcinoma cell line
Kristen D Brantley et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 27(6), 653-659 (2018-03-30)
Background: Expression of human paracrine hormones stanniocalcin 1 (STC1) and stanniocalcin 2 (STC2) may potentiate late breast cancer recurrence. We tested the hypothesis that expression of STC1 and STC2 in primary breast tumors is more strongly associated with late versus
Xin Ma et al.
Journal of translational medicine, 13, 56-56 (2015-03-06)
Although metastasis of clear cell renal cell carcinoma (ccRCC) is predominantly observed in late stage tumors, early stage metastasis of ccRCC can also be found with indefinite molecular mechanism, leading to inappropriate clinical decisions and poor prognosis. Stanniocalcin-1 (STC1) is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service