Skip to Content
Merck
All Photos(1)

Key Documents

HPA022148

Sigma-Aldrich

Anti-RAP1GAP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-GARNL4 antibody produced in rabbit, Anti-GTPase-activating Rap/Ran-GAP domain-like protein 4, Anti-Rap1 GTPase-activating protein 2, Anti-Rap1GAP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

MFGRKRSVSFGGFGWIDKTMLASLKVKKQELANSSDATLPDRPLSPPLTAPPTMKSSEFFEMLEKMQGIKLEEQKPGPQKNKDDYIPYPSIDEVVEKGGPYPQVI

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GARNL4(23108)

General description

Ras-related protein 1 GTPase activating protein 2 (RAP1GAP2) gene with 26 exons spanning 261 kilobases of genomic DNA, is mapped to human chromosome 17p13.3. RAP1GAP2 is a 715 amino acid protein found in platelets. The protein is characterized with a conserved central catalytic GAP domain, an N-terminal 14-3-3-binding site and a large C-terminal region of unknown function.

Immunogen

Rap1 GTPase-activating protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ras-related protein 1 GTPase activating protein 2 (RAP1GAP2) plays a vital role in regulation of platelet aggregation. It also controls the axon outgrowth in olfactory sensory neurons during early postnatal development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75630

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rap1gap2 regulates axon outgrowth in olfactory sensory neurons
Sadrian B, et al.
Molecular and Cellular Neurosciences, 50(3-4), 272-282 (2012)
Olga Neumüller et al.
Blood, 114(7), 1396-1404 (2009-06-17)
The small guanine-nucleotide-binding protein Rap1 plays a key role in platelet aggregation and hemostasis, and we recently identified Rap1GAP2 as the only GTPase-activating protein of Rap1 in platelets. In search of Rap1GAP2-associated proteins, we performed yeast-2-hybrid screening and found synaptotagmin-like
Jan Schultess et al.
Blood, 105(8), 3185-3192 (2005-01-06)
The Ras-like guanine-nucleotide-binding protein Rap1 controls integrin alpha(IIb)beta3 activity and platelet aggregation. Recently, we have found that Rap1 activation can be blocked by the nitric oxide/cyclic guanosine monophosphate (NO/cGMP) signaling pathway by type 1 cGMP-dependent protein kinase (cGKI). In search

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service