Skip to Content
Merck
All Photos(4)

Key Documents

HPA021234

Sigma-Aldrich

Anti-MRPL10 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-39S ribosomal protein L10, mitochondrial, Anti-L10mt, Anti-MRP-L10, Anti-MRP-L8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

NVALSAEDKLLMRHQLRKHKILMKVFPNQVLKPFLEDSKYQNLLPLFVGHNMLLVSEEPKVKEMVRILRTVPFLPLLGGC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL10(124995)

General description

MRPL10 (39S ribosomal protein L10, mitochondrial) is a protein in the 39 S ribosomal subunit of mitochondria. It exists in acetylated form.

Immunogen

39S ribosomal protein L10, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-MRPL10 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Acetylated MRPL10 (39S ribosomal protein L10, mitochondrial) causes high translational activity of mitochondrial ribosomes. It interacts with another protein MRPL12 in the ribosome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75720

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-Bo Li et al.
DNA and cell biology, 35(11), 680-690 (2016-10-12)
Mitochondrial ribosomal proteins are important for mitochondrial-encoded protein synthesis and mitochondrial function. In addition to their roles in mitoribosome assembly, several mitochondrial ribosome proteins are also implicated in cellular processes like cell cycle regulation, apoptosis, and mitochondrial homeostasis regulation. Here
Yongjie Yang et al.
The Journal of biological chemistry, 285(10), 7417-7429 (2010-01-01)
A member of the sirtuin family of NAD(+)-dependent deacetylases, SIRT3, is located in mammalian mitochondria and is important for regulation of mitochondrial metabolism, cell survival, and longevity. In this study, MRPL10 (mitochondrial ribosomal protein L10) was identified as the major
M A Piatyszek et al.
Nucleic acids research, 16(6), 2565-2583 (1988-03-25)
RNA binding properties of proteins from the large subunit of bovine mitochondrial ribosomes were studied using four different approaches: binding of radiolabeled RNA to western blotted proteins; disassembly of the intact 39 S ribosomal subunits with urea; binding of ribosomal
Jessica Nouws et al.
The Journal of biological chemistry, 291(2), 989-997 (2015-11-21)
To translate the 13 mtDNA-encoded mRNAs involved in oxidative phosphorylation (OXPHOS), mammalian mitochondria contain a dedicated set of ribosomes comprising rRNAs encoded by the mitochondrial genome and mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and imported into
Yulia V Surovtseva et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(44), 17921-17926 (2011-10-18)
Basal transcription of human mitochondrial DNA (mtDNA) in vitro requires the single-subunit, bacteriophage-related RNA polymerase, POLRMT, and transcription factor h-mtTFB2. This two-component system is activated differentially at mtDNA promoters by human mitochondrial transcription factor A (h-mtTFA). Mitochondrial ribosomal protein L7/L12

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service