Skip to Content
Merck
All Photos(6)

Key Documents

HPA020870

Sigma-Aldrich

Anti-GUCY1B3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GCS-beta-1, Anti-GCS-beta-3, Anti-Guanylate cyclase soluble subunit beta-1, Anti-Soluble guanylate cyclase small subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
6
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

HALELLVIRNYGPEVWEDIKKEAQLDEEGQFLVRIIYDDSKTYDLVAAASKVLNLNAGEILQMFGKMFFVFCQESGYDTILRVLGSNVREFLQNLDALHDHLATIYPGMRAPSFRCTDAEKGKGLILHYYSEREGLQDIVIGII

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GUCY1B3(2983)

General description

The gene GUCY1B3 (guanylate cyclase soluble subunit β-1) is mapped to human chromosome 4q31.3-q33. The protein localizes in the cytoplasm. GUCY1B3 has an amino-terminal regulatory domain, middle dimerization domain and carboxyl-terminal catalytic domain.

Immunogen

Guanylate cyclase soluble subunit beta-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GUCY1B3 (guanylate cyclase soluble subunit β-1) is a subunit of soluble guanylate cyclase. Soluble guanylate cyclase is a nitric oxide (NO) sensor. It is responsible for the synthesis of cGMP (3′,5′-cyclic guanosine monophosphate) from GTP (guanosine 5′-triphosphate) in response to NO. GUCY1B3 is up-regulated in mononuclear cells from humans suffering with erectile dysfunction.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72551

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles K Allerston et al.
PloS one, 8(3), e57644-e57644 (2013-03-19)
Soluble guanylate cyclase (sGC) catalyses the synthesis of cyclic GMP in response to nitric oxide. The enzyme is a heterodimer of homologous α and β subunits, each of which is composed of multiple domains. We present here crystal structures of
Jana Wobst et al.
Circulation journal : official journal of the Japanese Circulation Society, 79(3), 463-469 (2015-03-10)
Soluble guanylyl cyclase (sGC) is the physiological receptor for nitric oxide (NO) and NO-releasing drugs, and is a key enzyme in several cardiovascular signaling pathways. Its activation induces the synthesis of the second messenger cGMP. cGMP regulates the activity of
Swati Chauhan et al.
The Biochemical journal, 446(3), 445-453 (2012-06-14)
sGC (soluble guanylate cyclase) is the main mediator of NO signalling. Biochemical and physiological studies suggest that, besides NO, in vivo regulation of sGC involves direct interaction with other proteins. Using yeast two-hybrid screening, we identified that the multidomain LGN
Arnab Ghosh et al.
The Journal of biological chemistry, 289(22), 15259-15271 (2014-04-16)
The chaperone heat shock protein 90 (hsp90) associates with signaling proteins in cells including soluble guanylate cyclase (sGC). hsp90 associates with the heme-free (apo) sGC-β1 subunit and helps to drive heme insertion during maturation of sGC to its NO-responsive active
Fangfang Zhong et al.
Amino acids, 39(2), 399-408 (2010-01-12)
Soluble guanylate cyclase (sGC), as a nitric oxide (NO) sensor, is a critical heme-containing enzyme in NO-signaling pathway of eukaryotes. Human sGC is a heterodimeric hemoprotein, composed of a alpha-subunit (690 AA) and a heme-binding beta-subunit (619 AA). Upon NO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service