Skip to Content
Merck
All Photos(1)

Key Documents

HPA020553

Sigma-Aldrich

Anti-CIDEC antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Cell death activator CIDE-3, Anti-Cell death-inducing DFFA-like effector protein C, Anti-Fat-specific protein FSP27 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RDTLMLADKPFFLVLEEDGTTVETEEYFQALAGDTVFMVLQKGQKWQPPSEQGTRHPLSLSHKPAKKIDVARVT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CIDEC(63924)

General description

Cell death-inducing DFFA-like effector protein C (CIDEC) is localized on the surface of lipid droplets. The gene encoding it is localized on human chromosome 6.

Immunogen

Cell death activator CIDE-3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cell death-inducing DFFA-like effector protein C (CIDEC) has a role in the differentiation of adipocytes. It associates with 5′ adenosine monophosphate-activated protein kinase (AMPK) α subunit 1 and acts as a negative regulator of the protein complex. CIDEC is also involved in lipid storage and it modulates the size of lipid droplets. The protein has been linked to obesity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74259

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-resolution mapping of gene expression using association in an outbred mouse stock.
Ghazalpour A
PLoS Genetics, 4(8), e1000149-e1000149 (2008)
Seung Mi Lee et al.
Biochemical and biophysical research communications, 439(4), 564-569 (2013-09-13)
FSP27 (CIDE-3 in humans) plays critical roles in lipid metabolism and apoptosis and is known to be involved in regulation of lipid droplet (LD) size and lipid storage and apoptotic DNA fragmentation. Given that CIDE-containing proteins including FSP27 are associated
Z Q Wang et al.
International journal of obesity (2005), 34(9), 1355-1364 (2010-05-05)
It has been well documented that human adenovirus type 36 (Ad-36) is associated with obesity. However, the underlying molecular mechanism of Ad-36 inducing obesity remains unknown. We sought to investigate the effect of Ad-36 infection on Cidec, AMPK pathway and
Björn Morén et al.
Adipocyte, 9(1), 636-648 (2020-10-28)
Adipose tissue plays a major role in regulating whole-body energy metabolism. While the biochemical processes regulating storage and release of excess energy are well known, the temporal organization of these events is much less defined. In this study, we have
T Suami et al.
Carbohydrate research, 53(2), 239-246 (1977-02-01)
The aminocyclitol antibiotic neamine has been modified chemically by removing one or two hydroxyl groups from the 2-deoxystreptamine moiety to give 5- and 6- deoxyneamines (5 and 10), as well as 5,6-dideoxyneamine (15). Their antimicrobial activities were determined against several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service