Skip to Content
Merck
All Photos(2)

Key Documents

HPA019743

Sigma-Aldrich

Anti-UBN2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AC009220.10, Anti-Uncharacterized protein KIAA2030

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

SVTNQNVTPFGMLGGLVPVTMPFQFPLEIFGFGTDTAGVTTTSGSTSAAFHHSLTQNLLKGLQPGGAQHAATLSHSPLPAHLQQAFHDGGQSKGDTKLPRKS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBN2(254048)

General description

UBN2 (Ubinuclein 2) is a 12kDa protein consisting conserved HRD (Hpc2p-related domain) domain protein. Its mRNA expression has been found in variety of primary and transformed human cell types.

Immunogen

Uncharacterized protein KIAA2030 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

UBN2 (Ubinuclein 2) is a component of histone Chaperone Complex. In mammals, it plays a role in cell cycle regulation. It is involved in the various developmental processes as a part of Oct4 binding proteins including transcriptional regulation. It has been reported that UBN2 may have link with human developmental disorders or cancer. HRD domain of UBN2 binds to the WD repeats of human HIRA during cell senescence via chromatin-remodeling pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75035

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercedes Pardo et al.
Cell stem cell, 6(4), 382-395 (2010-04-07)
The transcription factor Oct4 is key in embryonic stem cell identity and reprogramming. Insight into its partners should illuminate how the pluripotent state is established and regulated. Here, we identify a considerably expanded set of Oct4-binding proteins in mouse embryonic
S Balaji et al.
Molecular bioSystems, 5(3), 269-275 (2009-02-20)
While histone chaperones have been intensely studied, the roles of components of the Hir-Asf1 histone chaperone complex such as Hir3p and Hpc2p are poorly understood. Using sensitive protein sequence profile analyses we investigated the evolution of these proteins and showed
Gowrishankar Banumathy et al.
Molecular and cellular biology, 29(3), 758-770 (2008-11-26)
Cellular senescence is an irreversible proliferation arrest, tumor suppression process and likely contributor to tissue aging. Senescence is often characterized by domains of facultative heterochromatin, called senescence-associated heterochromatin foci (SAHF), which repress expression of proliferation-promoting genes. Given its likely contribution
Trang Thi-Huynh Le et al.
American journal of cancer research, 12(1), 176-197 (2022-02-11)
Metastatic and castration-resistant disease is a fatal manifestation of prostate cancer (PCa). The mechanism through which resistance to androgen deprivation in PCa is developed remains largely unknown. Our understanding of the tumor microenvironment (TME) and key signaling pathways between tumors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service